EMAIL THIS PAGE TO A FRIEND
Tissue engineering. Part A
Chondrogenic potential of Slug-depleted human mesenchymal stem cells.
PMID 24712489
Abstract
The use of short interfering RNA (siRNA) in combination with stem cells and biocompatible scaffolds is a promising strategy in regenerative medicine. Our experimental strategy was to explore the possibility of forcing or guiding the chondrogenic differentiation of human mesenchymal stem cells (hMSCs) by knocking down a negative regulator of chondrogenesis, Slug transcription factor (TF), thus altering cell behavior. We found that TGFβ-driven chondrogenic differentiation of hMSCs cultured onto a hyaluronan-based scaffold, HYAFF(®)-11, was strengthened after cell exposure to siRNA against Slug. Slug silencing was effective in promoting the expression of chondrogenic markers, including Col2A1, aggrecan, Sox9, LEF1, and TRPS1. In addition, we confirmed that HYAFF-11 is a good scaffold candidate for hMSC use in tissue engineering applications, and showed that it is effective in sustaining TGFβ3 treatment associated with a specific gene silencing. Interestingly, preliminary results from the experimental model described here suggested that, even in the absence of differentiation supplements, Slug silencing showed a pro-chondrogenic effect, highlighting both its potential use as an alternative to TGFβ treatment, and the critical role of the Slug TF in determining the fate of hMSCs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N1665 5′-Nucleotidase human, recombinant, expressed in CHO cells, vial of 6-12 μg
SAB4700041 Anti-CD3, low endotoxin antibody, Mouse monoclonal, clone OKT3, purified immunoglobulin, buffered aqueous solution
SAB4300686
Anti-CD3E antibody produced in rabbit, affinity isolated antibody
CKOZFN15555 CompoZr® Knockout ZFN Kit, Human NT5E (NM_002526)
CKOZFND15555 CompoZr® Knockout ZFN Kit, ZFN plasmid only, Human NT5E (NM_002526)
EHU023471 MISSION® esiRNA, esiRNA human NT5E (esiRNA1)
SIHP0370 MISSION® Human Phosphatase, NT5E siRNA1, Nano Scale 250 pmol
SIHP0371 MISSION® Human Phosphatase, NT5E siRNA2, Nano Scale 250 pmol
SIHP0372 MISSION® Human Phosphatase, NT5E siRNA3, Nano Scale 250 pmol
C7048
Monoclonal Anti-CD3 antibody produced in mouse, clone UCHT-1, purified immunoglobulin, buffered aqueous solution
SAB4700040 Monoclonal Anti-CD3 antibody produced in mouse, clone OKT3, purified immunoglobulin, buffered aqueous solution
SAB4700044 Monoclonal Anti-CD3 antibody produced in mouse, clone MEM-57, purified immunoglobulin, buffered aqueous solution
SAB4700045 Monoclonal Anti-CD3 antibody produced in mouse, clone MEM-92, purified immunoglobulin, buffered aqueous solution
SAB4700050 Monoclonal Anti-CD3 antibody produced in mouse, clone APA1/1, purified immunoglobulin, buffered aqueous solution
WH0000916M4
Monoclonal Anti-CD3E antibody produced in mouse, clone 4C1, purified immunoglobulin, buffered aqueous solution
AMAB90876
Monoclonal Anti-CD3E antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1466, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90879
Monoclonal Anti-CD3E antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1497, purified immunoglobulin, buffered aqueous glycerol solution