EMAIL THIS PAGE TO A FRIEND
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Vicenin 2 isolated from Artemisia capillaris exhibited potent anti-glycation properties.
PMID 24713265
Abstract
Vicenin 2, isolated from a traditionally used medicinal plant Artemisia capillaris, is a 6,8-di-C-glucoside of apigenin which has been previously reported to possess a wide variety of pharmacological activities including antioxidant, anti-inflammatory, anti-cancer, and hepatoprotective. However, there have not been any reports concerning its anti-diabetic potential until now. Therefore, in the present study, we evaluated the anti-diabetic potential of vicenin 2 via α-glucosidase, protein tyrosine phosphatase 1B (PTP1B), rat lens aldose reductase (RLAR), and advanced glycation end products (AGE) formation inhibitory assays. Vicenin 2 strongly inhibited α-glucosidase, PTP1B, and RLAR in the corresponding assays. In addition, vicenin 2 inhibited the formation of both fluorescent AGE and nonfluorescent AGE, e.g., CML, as well as the level of fructosamine in glucose-fructose-induced bovine serum albumin (BSA) glycation. In the test system, vicenin 2 suppressed glycation-induced protein oxidation by attenuating the formation of protein carbonyl groups as well as by inhibiting the modification of protein thiol groups. Moreover, vicenin 2 was found to be a potent inhibitor of glycation-induced formation of amyloid cross-β structures in BSA. Taken together, vicenin 2 might be a useful lead for the development of multiple target-oriented therapeutic modalities for the treatment of diabetes and diabetes-associated complications.