EMAIL THIS PAGE TO A FRIEND
The American journal of pathology
M2 kupffer cells promote hepatocyte senescence: an IL-6-dependent protective mechanism against alcoholic liver disease.
PMID 24713392
Abstract
Alcoholic liver disease is a predominant cause of liver-related mortality in Western countries. The early steps of alcohol-induced steatosis and liver injury involve several mechanisms, including inflammation and oxidative stress. The inflammatory process is initiated by polarization of Kupffer cells toward a proinflammatory M1 phenotype, and we recently found that promoting anti-inflammatory M2 Kupffer cell polarization protects against alcohol-induced hepatocyte steatosis and apoptosis. Alcohol-induced oxidative stress is a potential trigger of senescence, and senescent cells exhibit characteristic functional resistance to apoptosis. We sought to evaluate induction of hepatocyte senescence as an early protective mechanism against alcoholic liver disease. Combining in vivo and in vitro studies, we show that M2 macrophages trigger hepatocyte senescence and enhance alcohol-induced hepatocyte senescence, as indicated by increased β-galactosidase activity, elevated CDKN1A mRNA expression, and induction of nuclear p21. We identify IL-6 as the mediator of M2-induced hepatocyte senescence. Senescent hepatocytes display characteristic resistance to apoptosis but also to steatosis, thus arguing for an early protective effect against alcoholic liver disease. These findings further suggest that pharmacologic interventions targeting M2 polarization during the early stages of alcoholic liver disease may represent an attractive strategy for the limitation of inflammation, hepatocyte apoptosis, and steatosis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2501669
Anti-CASP3 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA002643
Anti-CASP3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV00021
Anti-CASP3 antibody produced in rabbit, IgG fraction of antiserum
SAB4503291
Anti-CASP3 antibody produced in rabbit, affinity isolated antibody
C9598 Anti-Caspase 3 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2105613
Anti-D4S234E, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB2101297
Anti-KRAS antibody produced in rabbit, affinity isolated antibody
HPA049830
Anti-KRAS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2107115
Anti-KRAS antibody produced in rabbit, affinity isolated antibody
SAB2700181
Anti-NS3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2502090
Anti-Nsg1 (N-terminal) antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA035775
Anti-NSG1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4501190
Anti-NSG1 antibody produced in rabbit, affinity isolated antibody
P1998 Anti-Perilipin A antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AV42057
Anti-PLIN antibody produced in rabbit, IgG fraction of antiserum
HPA024299
Anti-PLIN1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4503030
Anti-TCTP antibody produced in rabbit, affinity isolated antibody
R3400 Monoclonal Anti-c-K-Ras antibody produced in mouse, clone 234-4.2, purified immunoglobulin, buffered aqueous solution
WH0000836M2 Monoclonal Anti-CASP3 antibody produced in mouse, clone 4D3, purified immunoglobulin, buffered aqueous solution
C5737 Monoclonal Anti-Caspase 3 antibody produced in mouse, clone 84803.111, purified immunoglobulin, lyophilized powder
WH0027065M1
Monoclonal Anti-D4S234E antibody produced in mouse, clone 1C3, purified immunoglobulin, buffered aqueous solution
WH0003845M1
Monoclonal Anti-KRAS antibody produced in mouse, clone 3B10-2F2, purified immunoglobulin, buffered aqueous solution
SAB1404011
Monoclonal Anti-KRAS antibody produced in mouse, clone 4F3, purified immunoglobulin, buffered aqueous solution
SAB4100599
Monoclonal Anti-KRAS antibody produced in mouse, clone KRAS-A582, culture supernatant
SAB4100600
Monoclonal Anti-KRAS antibody produced in mouse, clone KRAS-B582, culture supernatant
SAB1402313
Monoclonal Anti-PLIN antibody produced in mouse, clone 4G1, purified immunoglobulin, buffered aqueous solution
WH0007178M1
Monoclonal Anti-TPT1 antibody produced in mouse, clone 3C7, purified immunoglobulin, buffered aqueous solution