EMAIL THIS PAGE TO A FRIEND
Biotechnology and applied biochemistry
Dicistronic expression of human proinsulin-protein A fusion in tobacco chloroplast.
PMID 24716841
Abstract
Different expression systems such as bacteria and mammalian cells have been used to produce pharmaceutical proteins. In recent years, the use of plants as bioreactors offers efficient and economical systems in recombinant protein production. Furthermore, because of the large number of plastid copies in plants, chloroplast engineering functions as an effective method to increase recombinant protein expression. Because the commercially available insulin for treatment does not contain C-peptide, which is of great importance for type 1 diabetic patients, the current study introduces the human proinsulin gene fused with protein A into the tobacco chloroplast genome using the biolistic method. To achieve homoplasmy, three rounds of selection and regeneration of transforming cells were performed on the medium that contained spectinomycin antibiotic and hormones. The PCR analysis indicated the presence of the proinsulin gene in transplastomic plants. The reverse-transcription PCR analysis confirmed the expression of the proinsulin-proteinxa0A fusion at the transcription level. Immunoblot assays of leaf-derived protein extracts confirmed that the target gene expression is up to 0.2% of the total soluble protein. Our study showed that protein A fusion is not as efficient as other reported fusions. The transplastomic plants were also confirmed for homoplasmy using Southern blot analysis.