EMAIL THIS PAGE TO A FRIEND
International journal of oncology
The mushroom Ganoderma lucidum suppresses breast-to-lung cancer metastasis through the inhibition of pro-invasive genes.
PMID 24718855
Abstract
Breast cancer metastasis is one of the major reasons for the high morbidity and mortality of breast cancer patients. In spite of surgical interventions, chemotherapy, radiation therapy and targeted therapy, some patients are considering alternative therapies with herbal/natural products. In the present study, we evaluated a well-characterized extract from the medicinal mushroom Ganodermaxa0lucidum (GLE) for its affects on tumor growth and breast-to-lung cancer metastasis. MDA-MB-231 human breast cancer cells were implanted into the mammary fat pads of nude mice. GLE (100xa0mg/kg/every other day) was administered to the mice by an oral gavage for 4xa0weeks, and tumor size was measured using microcalipers. Lung metastases were evaluated by hematoxylin and eosin (H&E) staining. Gene expression in MDA-MB-231 cells was determined by DNA microarray analysis and confirmed by quantitative PCR. Identified genes were silenced by siRNA, and cell migration was determined in Boyden chambers and by wound-healing assay. Although an oral administration of GLE only slightly suppressed the growth of large tumors, the same treatment significantly inhibited the number of breast-to-lung cancer metastases. GLE also downregulated the expression of genes associated with invasive behavior (HRAS, VIL2, S100A4, MCAM, I2PP2A andxa0FN1) in MDA-MB-231 cells. Gene silencing of HRAS, VIL2, S100A4, I2PP2A and FN1 by siRNA suppressed migration of MDA-MB‑231 cells. Our study suggests that an oral administration of GLE can inhibit breast-to-lung cancer metastases through the downregulation of genes responsible for cell invasiveness. The anti-metastatic benefits of GLE warrant further clinical studies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4300372
Anti-EZR (Ab-353) antibody produced in rabbit, affinity isolated antibody
SAB4300460
Anti-EZR (Ab-567) antibody produced in rabbit, affinity isolated antibody
SAB1406584
Anti-EZR antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA021616
Anti-EZR antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
E1281
Anti-Ezrin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4503077
Anti-Ezrin antibody produced in rabbit, affinity isolated antibody
SAB4503078
Anti-Ezrin antibody produced in rabbit, affinity isolated antibody
GW21015 Anti-HRAS antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1405964
Anti-HRAS antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1406108
Anti-MCAM antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA008848
Anti-MCAM antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2700812
Anti-MCAM antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701024
Anti-MCAM antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1406406
Anti-S100A4 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA007973
Anti-S100A4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
S0575
Anti-SET (56-70) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2104853
Anti-SET, (N-terminal) antibody produced in rabbit, affinity isolated antibody
EHU134211 MISSION® esiRNA, esiRNA human HRAS (esiRNA1)
EMU210991 MISSION® esiRNA, esiRNA targeting mouse Hras1
SAB1402391
Monoclonal Anti-EZR antibody produced in mouse, clone 1E11, purified immunoglobulin, buffered aqueous solution
E8897
Monoclonal Anti-Ezrin antibody produced in mouse, clone 3C12, ascites fluid
SAB4100537
Monoclonal Anti-HRAS antibody produced in mouse, clone HRAS-A485, culture supernatant
WH0006275M1
Monoclonal Anti-S100A4 antibody produced in mouse, clone 1F12-1G7, purified immunoglobulin, buffered aqueous solution
AMAB90598
Monoclonal Anti-S100A4 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0239, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90596
Monoclonal Anti-S100A4 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0237, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90599
Monoclonal Anti-S100A4 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0240, purified immunoglobulin, buffered aqueous glycerol solution
WH0006418M1
Monoclonal Anti-SET antibody produced in mouse, clone M1-F5, purified immunoglobulin, buffered aqueous solution
SAB4200479
Monoclonal Anti-SET antibody produced in mouse, ~1.0 mg/mL, clone SET51, purified immunoglobulin
SAB4200498
Monoclonal Anti-SET antibody produced in mouse, clone SET7, tissue culture supernatant