EMAIL THIS PAGE TO A FRIEND
Experimental hematology
Mobilization with granulocyte colony-stimulating factor blocks medullar erythropoiesis by depleting F4/80(+)VCAM1(+)CD169(+)ER-HR3(+)Ly6G(+) erythroid island macrophages in the mouse.
PMID 24721610
Abstract
Similarly to other tissues, the bone marrow contains subsets of resident tissue macrophages, which are essential to maintain bone formation, functional hematopoietic stem cell (HSC) niches, and erythropoiesis. Pharmacologic doses of granulocyte colony-stimulating factor (G-CSF) mobilize HSC in part by interfering with the HSC niche-supportive function of BM resident macrophages. Because bone marrow macrophages are key to both maintenance of HSC within their niche and erythropoiesis, we investigated the effect of mobilizing doses of G-CSF on erythropoiesis in mice. We now report that G-CSF blocks medullar erythropoiesis by depleting the erythroid island macrophages we identified as co-expressing F4/80, vascular cell adhesion molecule-1, CD169, Ly-6G, and the ER-HR3 erythroid island macrophage antigen. Both broad macrophage depletion, achieved by injecting clodronate-loaded liposomes, and selective depletion of CD169(+) macrophages, also concomitantly depleted F4/80(+)VCAM-1(+)CD169(+)ER-HR3(+)Ly-6G(+) erythroid island macrophages and blocked erythropoiesis. This more precise phenotypic definition of erythroid island macrophages will enable studies on their biology and function in normal settings and on diseases associated with anemia. Finally, this study further illustrates that macrophages are a potent relay of innate immunity and inflammation on bone, hematopoietic, and erythropoietic maintenance. Agents that affect these macrophages, such as G-CSF, are likely to affect these three processes concomitantly.