EMAIL THIS PAGE TO A FRIEND
Oncotarget
Prostate cancer-derived CCN3 induces M2 macrophage infiltration and contributes to angiogenesis in prostate cancer microenvironment.
PMID 24721786
Abstract
Tumor-associated macrophages (TAMs) are M2-polarized macrophages that infiltrate the tumor microenvironment and promote tumorigenesis. However, the mechanisms by which TAMs modulate prostate cancer (PCa) growth are poorly understood. Here, we found that expression of Nephroblastoma Overexpressed (NOV/CCN3) is upregulated in PCa cells and correlated with M2 macrophage infiltration. RAW264.7 macrophage migration was induced by conditioned media (CM) from various PCa cells in proportion to the cellular level of CCN3 expression and was inhibited by an anti-CCN3 neutralizing antibody. CCN3 and PCaCM treatment skewed RAW264.7 cell differentiation from an M1 phenotype to an M2 phenotype. PCa-derived CCN3 induced focal adhesion kinase (FAK)/Akt/NF-κB signaling in RAW264.7 cells, which resulted in VEGF expression and subsequently increased tube formation in endothelial progenitor cells. Finally, PCa-secreted CCN3 stimulated RAW264.7 cells and promoted angiogenesis in the chick chorioallantoic membrane assay (CAM), and increased tumor growth and tumor-associated angiogenesis in a PCa xenograft mouse model. Our results indicate that PCa-secreted CCN3 can recruit macrophages and skew their differentiation to an M2 phenotype. In turn, CCN3-stimulated macrophages contribute to VEGF-dependent angiogenesis. This study reveals a novel mechanism by which TAMs enhance PCa angiogenesis and identifies a potential therapeutic target for PCa.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B3773 Anti-Human IgG (Fc specific)−Biotin antibody, Mouse monoclonal, clone HP-6017, purified from hybridoma cell culture
HPA004114
Anti-MRC1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA045134
Anti-MRC1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1301855 ANTI-MRC1L1 (N-TERM) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1406243
Anti-PECAM1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1408616
Anti-PECAM1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
B5283 Anti-Rabbit IgG (γ-chain specific)−Biotin antibody, Mouse monoclonal, clone RG-96, purified from hybridoma cell culture
A1949 Anti-Rabbit IgG (γ-chain specific)–Peroxidase antibody, Mouse monoclonal, clone RG-96, purified from hybridoma cell culture
R2655
Anti-Rabbit IgG, Native antibody, Mouse monoclonal, clone RabT-50, purified from hybridoma cell culture
V6627 Anti-Vascular Endothelial Growth Factor antibody produced in goat, IgG fraction of antiserum, lyophilized powder
V1253 Anti-Vascular Endothelial Growth Factor antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB2502119
Anti-VEGFA antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB4700463
Monoclonal Anti-CD31 antibody produced in mouse, clone MEM-05, purified immunoglobulin, buffered aqueous solution
G2904 Monoclonal Anti-Goat/Sheep IgG antibody produced in mouse, clone GT-34, ascites fluid
A8062 Monoclonal Anti-Goat/Sheep IgG–Alkaline Phosphatase antibody produced in mouse, clone GT-34, purified immunoglobulin, buffered aqueous glycerol solution
B3148 Monoclonal Anti-Goat/Sheep IgG–Biotin antibody produced in mouse, clone GT-34, purified immunoglobulin, buffered aqueous solution
F4891 Monoclonal Anti-Goat/Sheep IgG−FITC antibody produced in mouse, clone GT-34, purified immunoglobulin, buffered aqueous solution
A9452 Monoclonal Anti-Goat/Sheep IgG–Peroxidase antibody produced in mouse, clone GT-34, purified immunoglobulin, lyophilized powder
I6135 Monoclonal Anti-Human IgG (Fab specific) antibody produced in mouse, clone GG-6, ascites fluid
I6260 Monoclonal Anti-Human IgG (Fc specific) antibody produced in mouse, clone GG-7, ascites fluid
I6760 Monoclonal Anti-Human IgG (Fc specific) antibody produced in mouse, clone HP-6017, ascites fluid
F5016 Monoclonal Anti-Human IgG (Fc specific)−FITC antibody produced in mouse, clone HP-6017, purified immunoglobulin, buffered aqueous solution
I5885 Monoclonal Anti-Human IgG (γ-chain specific) antibody produced in mouse, clone GG-5, ascites fluid
I9513 Monoclonal Anti-Human IgG2 (Fc specific) antibody produced in mouse, clone HP-6002, ascites fluid
I9888 Monoclonal Anti-Human IgG4 (pFc′ specific) antibody produced in mouse, clone HP-6023, ascites fluid
WH0004360M2
Monoclonal Anti-MRC1 antibody produced in mouse, clone 5C11, purified immunoglobulin, buffered aqueous solution
AMAB90746
Monoclonal Anti-MRC1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0387, purified immunoglobulin, buffered aqueous glycerol solution
WH0005175M1
Monoclonal Anti-PECAM1 antibody produced in mouse, clone 1D2-1A5, purified immunoglobulin, buffered aqueous solution
R1008 Monoclonal Anti-Rabbit IgG (γ-chain specific) antibody produced in mouse, clone RG-96, ascites fluid
A2556 Monoclonal Anti-Rabbit IgG (γ-chain specific)−Alkaline Phosphatase antibody produced in mouse, clone RG-96, purified immunoglobulin, buffered aqueous glycerol solution