PloS one
A systematic comparison of the anti-tumoural activity and toxicity of the three Adv-TKs.
PMID 24722669
Abstract
Adenovirus 5 vectors, known respectively as, the first generation, second generation and oncolytic adenovirus, have been studied extensively in preclinical and clinical trials. However, hitherto few systemic evaluations of the efficacy and toxicity of these adenoviral vectors that have reflected the vertical history of adenovirus based cancer gene therapy strategies have been undertaken. This study has chosen Adv-TK, the well-established adjuvant treatment in cancer, and compared its efficacy and safety with those of the two newly synthesized oncolytic adenovirus vectors encoding the HSV-TK gene, namely M7 and M8. The results obtained showed that systemic administration of 1×10(8) pfu M7 had an anti-tumour efficacy similar to that of 3×10(10) pfu Adv-TK whilst M8 performed even better. Furthermore, compared to Adv-TK, M7 and M8 reduced the incidence of metastases and substantially prolonged the survival time of the mice xenografted with human orthotopic gastric carcinomas with disseminated metastasis. Even more exciting, however, were the similar toxic and immune safety results obtained from the administration of high doses of M7 or M8 in comparison with Adv-TK in immunocompetent and permissive syrian hamster. The data here exhibit a comprehensive display of the efficacy and safety of the three mutants and provide evidence for the future preclinical use of the M7 and M8 viruses.