EMAIL THIS PAGE TO A FRIEND
Electrophoresis
Biomarker discovery for neuroendocrine cervical cancer.
PMID 24723343
Abstract
Neuroendocrine cervical cancer is an aggressive but rare form of cervical cancer. The majority of neuroendocrine cervical cancer patients present with advanced-stage diseases. However, the limited numbers of neuroendocrine tumor markers are insufficient for clinical purposes. Thus, we used a proteomic approach combining lysine labeling 2D-DIGE and MALDI-TOF MS to investigate the biomarkers for neuroendocrine cervical cancer. By analyzing the global proteome alteration between the neuroendocrine cervical cancer line (HM-1) and non-neuroendocrine cervical cancer lines (CaSki cells, ME-180 cells, and Hela cells), we identified 82 proteins exhibiting marked changes between HM-1 and CaSki cells, and between ME-180 and Hela cells. Several proteins involved in protein folding, cytoskeleton, transcription control, signal transduction, glycolysis, and redox regulation exhibited significant changes in abundance. Proteomic and immunoblot analyses indicated respective 49.88-fold and 25-fold increased levels of transgelin in HM-1 cells compared with that in other non-neuroendocrine cervical cancer cell lines, implying that transgelin is a biomarker for neuroendocrine cervical cancer. In summary, we used a comprehensive neuroendocrine/non-neuroendocrine cervical cancer model based proteomic approach for identifying neuroendocrine cervical cancer markers, which might contribute to the prognosis and diagnosis of neuroendocrine cervical cancer.