EMAIL THIS PAGE TO A FRIEND
The British journal of dermatology
Expression dynamics of CXCL12 and CXCR4 during the progression of mycosis fungoides.
PMID 24725174
Abstract
Mycosis fungoides (MF) classically presents from patch stage to plaque stage over a number of years and finally progresses to tumour stage with nodal or visceral involvement. The mechanism of progression remains incompletely elucidated. Chemokines and their receptors are known to be involved in disease mechanisms, with CXCL12 and CXCR4 playing a critical role in carcinogenesis, invasion and cancer cell migration in various carcinomas. To investigate the expression of CXCL12 and CXCR4 in different cutaneous stages of MF. Formalin-fixed, paraffin-embedded skin samples from 40 patients with MF (21 patch stage, 10 plaque stage, nine tumour stage) and 30 non-neoplastic control skin samples were analysed. CXCL12 and CXCR4 were assessed by quantitative reverse-transcription polymerase chain reaction and immunohistochemical staining. The expression level of mRNA for CXCL12 in plaque-stage MF was significantly higher than in control skin (P = 0.0035), or patch-stage (P = 0.0108) or tumour-stage disease (P = 0.0089). The CXCR4 mRNA expression level in plaque-stage disease was significantly higher than in control skin (P = 0.0090) or patch-stage disease (P = 0.0387). CXCL12- and CXCR4-positive cell rates in patch-stage and plaque-stage MF were significantly higher than those in control skin (P < 0.0001). CXCL12- and CXCR4-positive cell rates in tumour-stage MF were significantly lower than those in patch- and plaque-stage disease (P = 0.0274 and P = 0.0492, respectively). Our data suggest that neoplastic T cells in MF are exposed to the microenvironment, given the abundance of CXCL12 during its progression, and also that neoplastic T cells express CXCR4, especially in the pretumour stage. We reveal that the CXCL12-CXCR4 axis plays a critical role in MF progression.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4700041 Anti-CD3, low endotoxin antibody, Mouse monoclonal, clone OKT3, purified immunoglobulin, buffered aqueous solution
SAB4300686
Anti-CD3E antibody produced in rabbit, affinity isolated antibody
SAB1401062
Anti-CD68 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2700244
Anti-CD68 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA048982
Anti-CD68 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2107746
Anti-CD68 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB2107747
Anti-CD68 antibody produced in rabbit, affinity isolated antibody
SAB1412069 ANTI-CXCL12 antibody produced in mouse, clone 2E2, purified immunoglobulin, buffered aqueous solution
SAB1412070
ANTI-CXCL12 antibody produced in mouse, clone 4D8, purified immunoglobulin, buffered aqueous solution
SAB1412168
ANTI-CXCL12 antibody produced in mouse, clone 1B2, purified immunoglobulin, buffered aqueous solution
SAB1412169
ANTI-CXCL12 antibody produced in mouse, clone 2A1, purified immunoglobulin, buffered aqueous solution
SAB1412233
ANTI-CXCL12 antibody produced in mouse, clone 2B1, purified immunoglobulin, buffered aqueous solution
SAB3501069
Anti-CXCL12 antibody produced in rabbit, affinity isolated antibody
GW21075 Anti-CXCR4 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
C3116
Anti-CXCR4 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3500383
Anti-CXCR4 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
C7048
Monoclonal Anti-CD3 antibody produced in mouse, clone UCHT-1, purified immunoglobulin, buffered aqueous solution
SAB4700040 Monoclonal Anti-CD3 antibody produced in mouse, clone OKT3, purified immunoglobulin, buffered aqueous solution
SAB4700044 Monoclonal Anti-CD3 antibody produced in mouse, clone MEM-57, purified immunoglobulin, buffered aqueous solution
SAB4700045 Monoclonal Anti-CD3 antibody produced in mouse, clone MEM-92, purified immunoglobulin, buffered aqueous solution
SAB4700050 Monoclonal Anti-CD3 antibody produced in mouse, clone APA1/1, purified immunoglobulin, buffered aqueous solution
WH0000916M4
Monoclonal Anti-CD3E antibody produced in mouse, clone 4C1, purified immunoglobulin, buffered aqueous solution
AMAB90876
Monoclonal Anti-CD3E antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1466, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90879
Monoclonal Anti-CD3E antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1497, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90873
Monoclonal Anti-CD68 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1338, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90874
Monoclonal Anti-CD68 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1346, purified immunoglobulin, buffered aqueous glycerol solution
WH0006387M1
Monoclonal Anti-CXCL12 antibody produced in mouse, clone 1E5, purified immunoglobulin, buffered aqueous solution
SAB1404360
Monoclonal Anti-CXCL12 antibody produced in mouse, clone 1F10, purified immunoglobulin, buffered aqueous solution
C6598 Monoclonal Anti-CXCR4 (Fusin) antibody produced in mouse, clone 44716, purified immunoglobulin, lyophilized powder
WH0007852M2
Monoclonal Anti-CXCR4 antibody produced in mouse, clone 1F8, ascites fluid