EMAIL THIS PAGE TO A FRIEND
International journal of pharmaceutics
A comparative ex vivo drug permeation study of beta-blockers through porcine buccal mucosa.
PMID 24727142
Abstract
Apparent permeability coefficients (kp) of a series of beta-blockers: acebutolol, atenolol, labetalol, metoprolol, oxprenolol and propranolol, through porcine buccal mucosa were determined. The aim of the study was to determine the permeation parameters (apparent permeability coefficient, kp; flux, J; and lag time, TL) as a measure of the intrinsic permeability of porcine buccal mucosa to these drugs, in order to predict the efficacy of their possible administration through human buccal mucosa. A positive linear correlation was observed between the apparent permeability coefficient, kpand the partition coefficient, P. Oxprenolol and propranolol are the drugs that presented the highest values of kp: 0.3231×10(2) cm/h and 0.5666×10(2) cm/h, respectively. Multiple linear regression (MLR) using least square estimation was performed on the data set with logkpas dependent variable and the descriptors as predictor variables. The potential systemic capacity after a buccal administration was predicted by estimating the plasma concentrations at steady-stated (Css). Considering the entire process of permeation ex vivo, propranolol and oxprenolol would seem to be the best candidates for administration through the buccal mucosa.