EMAIL THIS PAGE TO A FRIEND
Journal of bacteriology
Arginine deiminase in Staphylococcus epidermidis functions to augment biofilm maturation through pH homeostasis.
PMID 24727224
Abstract
Allelic replacement mutants were constructed within arginine deiminase (arcA1 and arcA2) to assess the function of the arginine deiminase (ADI) pathway in organic acid resistance and biofilm formation of Staphylococcus epidermidis 1457. A growth-dependent acidification assay (pH ∼5.0 to ∼5.2) determined that strain 1457 devoid of arginine deiminase activity (1457 ΔADI) was significantly less viable than the wild type following depletion of glucose and in the presence of arginine. However, no difference in viability was noted for individual 1457 ΔarcA1 (native) or ΔarcA2 (arginine catabolic mobile element [ACME]-derived) mutants, suggesting that the native and ACME-derived ADIs are compensatory in S. epidermidis. Furthermore, flow cytometry and electron paramagnetic resonance spectroscopy results suggested that organic acid stress resulted in oxidative stress that could be partially rescued by the iron chelator dipyridyl. Collectively, these results suggest that formation of hydroxyl radicals is partially responsible for cell death via organic acid stress and that ADI-derived ammonia functions to counteract this acid stress. Finally, static biofilm assays determined that viability, ammonia synthesis, and pH were reduced in strain 1457 ΔADI following 120 h of growth in comparison to strain 1457 and the arcA1 and arcA2 single mutants. It is hypothesized that ammonia synthesis via the ADI pathway is important to reduce pH stress in specific microniches that contain high concentrations of organic acids.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1270000
Arginine, European Pharmacopoeia (EP) Reference Standard
C6H14N4O2
C3290000
Cysteine hydrochloride monohydrate, European Pharmacopoeia (EP) Reference Standard
C3H7NO2S · HCl · H2O
C121800
L-Cysteine hydrochloride hydrate, 99%
C3H7NO2S · HCl · xH2O
C7880
L-Cysteine hydrochloride monohydrate, reagent grade, ≥98% (TLC)
C3H7NO2S · HCl · H2O
C6852
L-Cysteine hydrochloride monohydrate, from non-animal source, cell culture tested, meets EP, USP testing specifications
C3H7NO2S · HCl · H2O
C5610
L-Cysteine hydrochloride monohydrate, PharmaGrade, Ajinomoto, EP, USP, manufactured under appropriate GMP controls for Pharma or Biopharmaceutical production, suitable for cell culture
C3H7NO2S · HCl · H2O
30122
L-Cysteine hydrochloride monohydrate, tested according to Ph.Eur.
C3H7NO2S · HCl · H2O
W779989
L-Cysteine hydrochloride monohydrate, FG, produced by Wacker Chemie AG, Burghausen, Germany, Food, 20 mesh
C3H7NO2S · HCl · H2O
W779865
L-Cysteine hydrochloride monohydrate, 99%, FG, produced by Wacker Chemie AG, Burghausen, Germany
C3H7NO2S · HCl · H2O
PHR1102
L-Cysteine hydrochloride monohydrate, Pharmaceutical Secondary Standard; Certified Reference Material
C3H7NO2S · HCl · H2O
30129
L-Cysteine hydrochloride monohydrate, BioUltra, ≥99.0% (RT)
C3H7NO2S · HCl · H2O
30130
L-Cysteine hydrochloride monohydrate, ≥99.0% (RT)
C3H7NO2S · HCl · H2O
778451
L-Cysteine hydrochloride monohydrate, Produced by Wacker Chemie AG, Burghausen, Germany, Life Science, 98.5-101.0% (Ph.Eur (2.2.20))
C3H7NO2S · HCl · H2O