EMAIL THIS PAGE TO A FRIEND
International journal of oncology
The expression and phosphorylation of ezrin and merlin in human pancreatic cancer.
PMID 24728215
Abstract
Pancreatic carcinoma is the most common pancreatic malignancy and is associated with a very poor prognosis. Therefore, new prognostic factors and new treatment strategies are clearly needed. In this study, we retrospectively studied the levels of phosphorylated ezrin in 19 patients with pancreatic carcinoma by immunohistochemical analysis and determined the correlation between protein expression, clinicopathological characteristics and prognosis in pancreatic adenocarcinoma. We also characterized the phenotype of the overexpression of wild-type and phosphorylated ezrin and merlin in human pancreatic cancer cell lines. A significant correlation between the levels of phosphorylated ezrinxa0353 and ezrinxa0567 and the stage of pancreatic cancer was observed. Moreover, Kaplan-Meier analysis revealed that patients with high levels of phosphorylated ezrin had a significantly poorer survival rate (P<0.05). In addition, the overexpression of wild-type merlin or ezrin inhibited cell proliferation, migration and adhesion. However, the overexpression of T567D ezrin, a mutant that mimics permanent phosphorylation, promoted the proliferation, adhesion and migration of the pancreatic adenocarcinoma cell line SW1990. The overexpression of S518D merlin inhibited the growth of SW1990 and did not affect migration or adhesion. These results suggest that the phosphorylation of ezrin may contribute to the progression of pancreatic carcinoma and that the level of phosphorylated ezrin may serve as an adverse prognostic factor for pancreatic carcinoma.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4300372
Anti-EZR (Ab-353) antibody produced in rabbit, affinity isolated antibody
SAB4300460
Anti-EZR (Ab-567) antibody produced in rabbit, affinity isolated antibody
SAB1406584
Anti-EZR antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA021616
Anti-EZR antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
E1281
Anti-Ezrin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4503077
Anti-Ezrin antibody produced in rabbit, affinity isolated antibody
SAB4503078
Anti-Ezrin antibody produced in rabbit, affinity isolated antibody
SAB4300162
Anti-phospho-EZR (pThr567) antibody produced in rabbit, affinity isolated antibody
SAB4300049
Anti-phospho-EZR (pTyr353) antibody produced in rabbit, affinity isolated antibody
SAB4504401
Anti-phospho-Ezrin (pThr566) antibody produced in rabbit, affinity isolated antibody
SAB4504581
Anti-phospho-Ezrin (pTyr146) antibody produced in rabbit, affinity isolated antibody
SAB4504402
Anti-phospho-Ezrin (pTyr353) antibody produced in rabbit, affinity isolated antibody
SAB4504582
Anti-phospho-Ezrin (pTyr478) antibody produced in rabbit, affinity isolated antibody
SAB1402391
Monoclonal Anti-EZR antibody produced in mouse, clone 1E11, purified immunoglobulin, buffered aqueous solution