PloS one
Substrate modulation of fatty acid effects on energization and respiration of kidney proximal tubules during hypoxia/reoxygenation.
PMID 24728405
Abstract
Kidney proximal tubules subjected to hypoxia/reoxygenation develop a nonesterified fatty acid-induced energetic deficit characterized by persistent partial mitochondrial deenergization that can be prevented and reversed by citric acid cycle substrates. To further assess the role of competition between fatty acids and substrates on inner membrane substrate carriers in the deenergization and the contribution to deenergization of fatty acid effects on respiratory function, digitonin-permeabilized rabbit and mouse tubules were studied using either addition of exogenous oleate after control normoxic incubation or increases of endogenous fatty acids produced by hypoxia/reoxygenation. The results demonstrated major effects of matrix oxaloacetate accumulation on succinate-supported energization and respiration and their modification by fatty acids. Improvements of energization in the presence of fatty acids by glutamate were shown to result predominantly from lowering matrix oxaloacetate rather than from amelioration of transmembrane cycling of fatty acids and uncoupling. Mouse tubules had 2.5 fold higher rates of succinate utilization, which resulted in stronger effects of oxaloacetate accumulation than rabbit tubules. Hypoxia/reoxygenation induced respiratory inhibition that was more severe for complex I-dependent substrates. Fatty acids themselves did not acutely contribute to this respiratory inhibition, but lowering them during 60 min. reoxygenation to allow recovery of ATP during that period alleviated it. These data clarify the basis for the nonesterified fatty acid-induced mitochondrial energetic deficit in kidney proximal tubules that impairs structural and functional recovery and provide insight into interactions that need to be considered in the design of substrate-based interventions to improve mitochondrial function.