EMAIL THIS PAGE TO A FRIEND
Journal of nanoscience and nanotechnology
Biological applications of gold nanoparticles.
PMID 24730267
Abstract
This article reviews some of the recent biological applications of gold nanoparticles (GNPs) which have been discovered lately by individual studies all around the world. GNPs have emerged as a promising candidates for various biological applications due to their unique physical properties (size and shape dependent), excellent biocompatibility, facile synthesis, ease of bioconjugation, etc. This review starts with a brief introduction about nanotechnology followed by an insight into the history, emergence, and enhanced properties of various gold nanostructures, which form the basis for their numerous biomedical applications. In addition, a brief overview on some of the commonly used fabrication techniques for synthesizing GNPs is also discussed. Finally, a miscellany of the latest biological applications of GNPs, such as cancer diagnostics and therapy, biological probes, drug delivery, gene delivery, vaccine preparation, brain implants, artificial skin, sterilization system, and improving electrical signaling in the heart, published in different articles in reputed journals are highlighted.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
326542
Gold, beads, 1-6 mm, 99.999% trace metals basis
Au
373168
Gold, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.99% trace metals basis
Au
373176
Gold, evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.99% trace metals basis
Au
373184
Gold, evaporation slug, diam. × L 0.3 cm × 0.6 cm, 99.99% trace metals basis
Au
265829
Gold, foil, thickness 0.5 mm, 99.99% trace metals basis
Au
349240
Gold, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Au
326496
Gold, foil, thickness 0.127 mm, 99.99% trace metals basis
Au
265810
Gold, foil, thickness 0.1 mm, 99.99% trace metals basis
Au
349275
Gold, foil, thickness 0.05 mm, 99.99% trace metals basis
Au
268461
Gold, foil, thickness 0.025 mm, 99.99% trace metals basis
Au
349267
Gold, foil, thickness 0.025 mm, ≥99.9% trace metals basis
Au
255734
Gold, powder, <850 μm, ≥99.99% trace metals basis
Au
265772
Gold, powder, <45 μm, 99.99% trace metals basis
Au
718424
Gold, powder, size 10-20 μm
Au
326585
Gold, powder, <10 μm, ≥99.9% trace metals basis
Au
636347
Gold, nanopowder, <100 nm particle size, 99.9% trace metals basis
Au
265837
Gold, rod, diam. 3.0 mm, 99.99% trace metals basis
Au
265802
Gold, wire, diam. 2.0 mm, 99.99% trace metals basis
Au
349291
Gold, wire, diam. 1.5 mm, 99.999% trace metals basis
Au
349305
Gold, wire, diam. 1.0 mm, 99.997% trace metals basis
Au
265799
Gold, wire, diam. 1.0 mm, 99.99% trace metals basis
Au
349313
Gold, wire, diam. 0.5 mm, 99.999% trace metals basis
Au
310980
Gold, wire, diam. 0.5 mm, 99.99% trace metals basis
Au
265780
Gold, wire, diam. 0.25 mm, 99.99% trace metals basis
Au
326534
Gold, wire, diam. 0.25 mm, ≥99.9% trace metals basis
Au
326526
Gold, wire, diam. 0.127 mm, 99.99% trace metals basis
Au
349283
Gold, wire, diam. 0.1 mm, 99.99% trace metals basis
Au
GF43139538
Gold, insulated wire, 5m, conductor diameter 0.025mm, insulation thickness 0.005mm, polyester insulation, 99.99%
Au
GF92307922
Gold, insulated wire, 5m, conductor diameter 0.05mm, insulation thickness 0.007mm, polyester insulation, 99.99%
Au
GF11444611
Gold, insulated wire, 5m, conductor diameter 0.05mm, insulation thickness 0.007mm, polyimide insulation, 99.99%
Au