EMAIL THIS PAGE TO A FRIEND
Journal of drug targeting
Enhancement of antifungal activity by integrin-targeting of branched histidine rich peptides.
PMID 24731059
Abstract
The treatment of invasive candidiasis associated with growing numbers of immunocompromised patients remains a major challenge complicated by increasing drug resistance. A novel class of branched histidine-lysine (bHK) peptides has promising antifungal activity, and exhibits a mechanism similar to natural histatins, and thus may avoid drug resistance. The present studies evaluate ligand targeting of bHK peptides to fungal surface integrins by determining whether a cyclic RGD (cRGD) peptide with a large PEG linker could enhance bHK peptide antifungal activity. Whereas conjugates containing only the PEG linker reduced bHK peptide activity, conjugates with the cRGD-PEG ligand resulted in marked enhancement of activity against Candida albicans. This study provides the first demonstration of benefit from ligand targeting of antifungal agents to fungal surface receptors.