EMAIL THIS PAGE TO A FRIEND
Molecular immunology
Tenocyte activation and regulation of complement factors in response to in vitro cell injury.
PMID 24732065
Abstract
Inferior tendon healing can lead to scarring and tendinopathy. The role of complement in tendon healing is still unclear. The aim of this study was to understand tenocytes response to mechanical injury and whether complement is regulated by injury. Tenocytes were injured using an optimized automated scratch assay model. Using a self-assembled plotter system, 50 parallel lines of injury were created in a 6 cm diameter tenocyte cell layer. Tenocytes mitotic activity and survival post injury was assessed using FDA/ethidiumbromide assay. Furthermore, this injury model was combined with stimulation of the tenocytes with the complement split fragment C3a. Gene expression of C3aR, C5aR (CD88), CD46, CD55, tumor necrosis factor (TNF)α, interleukin (IL)-1β, matrix metalloproteinase (MMP)-1 was analyzed. Immunolabeling for C5aR and CD55 was performed. An enhanced mitotic activity and some dead cells were detected in the vicinity of the scratches. Gene expression of the C3aR was suppressed after 4 h but induced after 24 h post injury. C5aR was down-regulated at 24 h, CD46 and CD55 were induced at 24 h in response to injury and CD55 was also elevated at 4 h. MMP-1 was upregulated by injury but both proinflammatory cytokines remained mainly unaffected. Combination of injury with C3a stimulation led to an enhanced C3aR, CD55 and TNFα gene expression. According to the gene expression data, the protein expression of C5aR was reduced and that of CD55 induced. In summary, a specific response of complement regulation was found in mechanically injured tenocytes which may be involved in healing responses.