PloS one
Zinc regulates the acute phase response and serum amyloid A production in response to sepsis through JAK-STAT3 signaling.
PMID 24732911
Abstract
Sepsis rapidly activates the host inflammatory response and acute phase response. Severe sepsis, complicated by multiple organ failure, is associated with overwhelming inflammation and high mortality. We previously observed that zinc (Zn) deficiency significantly increases mortality in a mouse model of polymicrobial sepsis due to over-activation of the inflammatory response. In order to identify potential mechanisms that account for Zn-responsive effects, we generated whole exome expression profiles from the lung tissue of septic mice that were maintained on Zn modified diets. Based on systems analysis, we observed that Zn deficiency enhances the acute phase response and particularly the JAK-STAT3 pathway, resulting in increased serum amyloid A production. In vitro studies of primary hepatocytes and HepG2 cells substantiated that Zn-deficiency augments serum amyloid A production through up-regulation of the JAK-STAT3 and NF-κB pathways. In contrast, Zn inhibited STAT3 activation through the up-regulation of SHP1 activity. Collectively, these findings demonstrate that Zn deficiency enhances the acute phase response through up-regulation of the JAK-STAT3 pathway, thereby perpetuating increased inflammation that may lead to increased morbidity and mortality in response to sepsis.