EMAIL THIS PAGE TO A FRIEND
Psychopharmacology
Phosphorylation of hypothalamic AMPK on serine(485/491) related to sustained weight loss by alpha-lipoic acid in mice treated with olanzapine.
PMID 24733236
Abstract
Alpha-lipoic acid (ALA) was shown to suppress atypical antipsychotic drug (AAPD)-induced weight gain. However, its mode of action has remained unidentified. We aimed to identify mechanisms underlying anti-obesity effects of ALA in mice treated with olanzapine. We compared body weight and food intake among vehicle-, olanzapine-, and olanzapine plus ALA-treated mice, and measured hypothalamic AMP-activated protein kinase (AMPK) activity by detecting levels of Thr(172) and Ser(485/491) phosphorylation, which indicate activation and inhibition of AMPK, respectively. Body weights were increased by olanzapine in parallel with increased levels of Thr(172) phosphorylation of hypothalamic AMPK. Initially increased rate of weight gain was diminished as Thr(172) phosphorylation levels were decreased to control levels after 10xa0days of olanzapine treatment. ALA successfully not only prevented olanzapine-induced weight gain but also induced additional weight loss even relative to control levels throughout the treatment period. During the initial stage, ALA's action was indicated by both suppression of olanzapine-induced Thr(172) phosphorylation and an increase in Ser(485/491) phosphorylation levels. However, in the later stage when no more increases in Thr(172) phosphorylation and weight gain by olanzapine were observed, ALA's action was only indicated by increased levels of Ser(485/491) phosphorylation. Our data suggest that anti-obesity effects of ALA may be related to modulation of both Ser(485/491) phosphorylation and Thr(172) phosphorylation of hypothalamic AMPK, while olanzapine-induced weight gain may be only associated with increase in Thr(172) phosphorylation. This might be an important mechanistic clue for thexa0future development of anti-obesity drugs beyond control of AAPD-induced weight gain.