PloS one
Renal involvement in non-Hodgkin lymphoma: proven by renal biopsy.
PMID 24733356
Abstract
To determine the spectrum of renal lesions in patients with kidney involvement in non-Hodgkin's lymphoma (NHL) by renal biopsy. The clinical features and histological findings at the time of the renal biopsy were assessed for each patient. We identified 20 patients with NHL and renal involvement, and the diagnosis of NHL was established following the kidney biopsy in 18 (90%) patients. The types of NHL include the following: chronic lymphocytic leukemia/small lymphocytic lymphoma (n = 8), diffuse large B-cell lymphoma (n = 4), T/NK cell lymphoma (n = 3), lymphoplasmacytic lymphoma (n = 2), cutaneous T-cell lymphoma (n = 1), mucosa-associated lymphoid tissue lymphoma (n = 1) and mantle cell lymphoma (n = 1). All presented with proteinuria, and 15 patients had impaired renal function. The pathological findings included (1) membranoproliferative glomerulonephritis-like pattern in seven patients; (2) crescent glomerulonephritis in four; (3) minimal-change disease in three, and glomeruli without specific pathological abnormalities in three; (4) intraglomerular large B-cell lymphoma in one; (5) intracapillary monoclonal IgM deposits in one; (6) primary diffuse large B-cell lymphoma of the kidneys in one; and (7) lymphoma infiltration of the kidney in eight patients. A wide spectrum of renal lesions can be observed in patients with NHL, and NHL may be first proven by renal biopsies for evaluation of kidney injury or proteinuria. Renal biopsy is necessary to establish the underlying cause of renal involvement in NHL.