EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Common metal of copper(0) as an efficient catalyst for preparation of nitriles and imines by controlling additives.
PMID 24733424
Abstract
A novel, efficient, convenient and environmentally friendly approach for the synthesis of nitriles and imines from primary amines has been developed. Using commercially available red copper as the catalyst, ammonium bromide as the co-catalyst and molecular oxygen as the sole oxidant, nitriles and imines can be afforded in high yields through benzylic oxidation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
479152
4-Bromobenzylamine, 96%
C7H8BrN