EMAIL THIS PAGE TO A FRIEND
Annals of the New York Academy of Sciences
Translational research investigations on ATP7A: an important human copper ATPase.
PMID 24735419
Abstract
In more than 40 years since copper deficiency was delineated in pediatric subjects with Menkes disease, remarkable advances in our understanding of the clinical, biochemical, and molecular aspects of the human copper transporter ATP7A have emerged. Mutations in the gene encoding this multitasking molecule are now implicated in at least two other distinctive phenotypes: occipital horn syndrome and ATP7A-related isolated distal motor neuropathy. Several other novel inherited disorders of copper metabolism have been identified in the past several years, aided by advances in human gene mapping and automated DNA sequencing. In this paper, I review the history and evolution of our understanding of disorders caused by impaired ATP7A function, and outline future challenges.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF14058705
Copper, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.999%
Cu
GF71667366
Copper, wire reel, 1000m, diameter 0.04mm, annealed, 99.9%
Cu
GF21709650
Copper, wire reel, 1000m, diameter 0.04mm, hard, 99.9%
Cu
GF99677165
Copper, wire reel, 1000m, diameter 0.05mm, annealed, 99.9%
Cu
GF08494577
Copper, wire reel, 1000m, diameter 0.075mm, annealed, 99.9%
Cu
GF66965956
Copper, wire reel, 1000m, diameter 0.125mm, annealed, 99.9%
Cu
GF22319872
Copper, wire reel, 1000m, diameter 0.15mm, annealed, silver plated, 99.9%
Cu
GF27350287
Copper, wire reel, 1000m, diameter 0.1mm, annealed, 99.9%
Cu
GF63655780
Copper, wire reel, 1000m, diameter 0.25mm, annealed, 99.9%
Cu
GF47171033
Copper, wire reel, 100m, diameter 0.025mm, as drawn, 99.99+%
Cu
GF18312791
Copper, wire reel, 100m, diameter 0.02mm, hard, 99.99+%
Cu
GF89712333
Copper, wire reel, 100m, diameter 0.5mm, as drawn, 99.98+%
Cu
GF93417105
Copper, wire reel, 100m, diameter 1.0mm, as drawn, 99.98+%
Cu
GF64298829
Copper, wire reel, 100m, diameter 0.02mm, annealed, 99.99%
Cu
GF86528155
Copper, wire reel, 100m, diameter 0.05mm, annealed, 99.9%
Cu
GF39045090
Copper, wire reel, 100m, diameter 0.075mm, annealed, 99.9%
Cu
GF71409089
Copper, wire reel, 100m, diameter 0.125mm, annealed, 99.9%
Cu
GF42860651
Copper, wire reel, 100m, diameter 0.15mm, annealed, silver plated, 99.9%
Cu
GF52128008
Copper, wire reel, 100m, diameter 0.1mm, annealed, 99.9%
Cu
GF69188079
Copper, wire reel, 100m, diameter 0.25mm, annealed, 99.9%
Cu
GF98066054
Copper, wire reel, 100m, diameter 0.5mm, annealed, 99.9%
Cu
GF81594399
Copper, wire reel, 100m, diameter 0.5mm, annealed, silver plated, 99.9%
Cu
GF82471620
Copper, wire reel, 100m, diameter 1.0mm, annealed, 99.9%
Cu
GF83100619
Copper, wire reel, 10m, diameter 0.025mm, as drawn, 99.99+%
Cu
GF80026770
Copper, wire reel, 10m, diameter 0.02mm, hard, 99.99+%
Cu
GF74431324
Copper, wire reel, 10m, diameter 0.05mm, as drawn, 99.99+%
Cu
GF81301609
Copper, wire reel, 10m, diameter 0.75mm, as drawn, 99.98+%
Cu
GF86675439
Copper, wire reel, 10m, diameter 0.01mm, hard, 99.99%
Cu
GF77356109
Copper, wire reel, 10m, diameter 0.02mm, annealed, 99.99%
Cu
GF56973257
Copper, wire reel, 2000m, diameter 0.25mm, annealed, 99.9%
Cu