PloS one
Tissue inhibitor of metalloproteinase-3 knockout mice exhibit enhanced energy expenditure through thermogenesis.
PMID 24736588
Abstract
Tissue inhibitors of metalloproteinases (TIMPs) regulate matrix metalloproteinase activity and maintain extracellular matrix homeostasis. Although TIMP-3 has multiple functions (e.g., apoptosis, inhibition of VEGF binding to VEGF receptor, and inhibition of TNFα converting enzyme), its roles in thermogenesis and metabolism, which influence energy expenditure and can lead to the development of metabolic disorders when dysregulated, are poorly understood. This study aimed to determine whether TIMP-3 is implicated in metabolism by analyzing TIMP-3 knockout (KO) mice. TIMP-3 KO mice had higher body temperature, oxygen consumption, and carbon dioxide production than wild-type (WT) mice, although there were no differences in food intake and locomotor activity. These results suggest that metabolism is enhanced in TIMP-3 KO mice. Real-time PCR analysis showed that the expression of PPAR-δ, UCP-2, NRF-1 and NRF-2 in soleus muscle, and PGC-1α and UCP-2 in gastrocnemius muscle, was higher in TIMP-3 KO mice than in WT mice, suggesting that TIMP-3 deficiency may increase mitochondrial activity. When exposed to cold for 8 hours to induce thermogenesis, TIMP-3 KO mice had a higher body temperature than WT mice. In the treadmill test, oxygen consumption and carbon dioxide production were higher in TIMP-3 KO mice both before and after starting exercise, and the difference was more pronounced after starting exercise. Our findings suggest that TIMP-3 KO mice exhibit enhanced metabolism, as reflected by a higher body temperature than WT mice, possibly due to increased mitochondrial activity. Given that TIMP-3 deficiency increases energy expenditure, TIMP-3 may present a novel therapeutic target for preventing metabolic disorders.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
56454
1-Thioglycerol, PharmaGrade, USP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.
C3H8O2S
M1753
1-Thioglycerol, ≥97%
C3H8O2S
M6145
1-Thioglycerol, liquid, BioReagent, suitable for cell culture, ≥97% (titration)
C3H8O2S
M2172
1-Thioglycerol, BioXtra, ≥97%
C3H8O2S
88640
1-Thioglycerol, ≥99.0% (GC)
C3H8O2S
88642
1-Thioglycerol, dist., ≥98.0% (GC)
C3H8O2S
SAB2500268
Anti-COX4I1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1405644
Anti-COX4I1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2701802
Anti-cox4i1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA002485
Anti-COX4I1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1401897
Anti-COX4I2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2101653
Anti-NRF1 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2101654
Anti-NRF1 (ab2) antibody produced in rabbit, affinity isolated antibody
AV38543
Anti-NRF1 antibody produced in rabbit, IgG fraction of antiserum
HPA029329
Anti-NRF1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1400293
Anti-UCP2 antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
SAB1409788
Anti-UCP2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1409789
Anti-UCP2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1411380
Anti-UCP2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB5300321
Monoclonal Anti-COX4I1 antibody produced in mouse, clone 6B3, ascites fluid
SAB1402152
Monoclonal Anti-COX4I1 antibody produced in mouse, clone 4A10, purified immunoglobulin, buffered aqueous solution
WH0084701M1
Monoclonal Anti-COX4I2 antibody produced in mouse, clone 1F2, purified immunoglobulin, buffered aqueous solution
SAB1404147
Monoclonal Anti-NRF1 antibody produced in mouse, clone 1B9, purified immunoglobulin, buffered aqueous solution