EMAIL THIS PAGE TO A FRIEND
Carcinogenesis
Endothelial cell-derived fibronectin extra domain A promotes colorectal cancer metastasis via inducing epithelial-mesenchymal transition.
PMID 24743511
Abstract
Recent evidence has been suggesting the important roles of endothelial cells (ECs) involved in the pathogenesis of several cancers, including colorectal carcinomas (CRCs), but the underlying mechanism remains elusive. We have demonstrated previously that CRC-derived fibronectin extra domain A (EDA) promotes vasculogenesis, tumorigenesis and metastasis of CRCs. At the current study, we showed that EC-secreted EDA promotes the metastatic capacity CRC cells via inducing an epithelial-mesenchymal transition. In vitro and in vivo experiments showed that EC-secreted EDA, via the interaction with integrin α9β1 on neighboring CRC cells, leads to the activation of focal adhesion kinase as well as Rac signalings, thus strengthens the polarity of cytoskeleton and promotes the invasion capacity of CRC cells. Furthermore, Erk signaling pathway was revealed to critically mediate the effect of EC-derived EDA on CRC cells. Our findings reveal a novel oncogenic role of ECs in promoting CRC malignancy through secreting EDA.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1405763
Anti-EPHB1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1412026
ANTI-EPHB2 antibody produced in mouse, clone 1F10, ascites fluid, buffered aqueous solution
T6528
Anti-Mouse IgG (Fab specific)–TRITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
T7782 Anti-Mouse IgG (Fab specific)–TRITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB4504583
Anti-phospho-FAK (pSer843) antibody produced in rabbit, affinity isolated antibody
SAB4504180
Anti-phospho-FAK (pSer910) antibody produced in rabbit, affinity isolated antibody
F7926 Anti-phospho-FAK (pTyr397) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
SAB4504181
Anti-phospho-FAK (pTyr397) antibody produced in rabbit, affinity isolated antibody
SAB4504403
Anti-phospho-FAK (pTyr397) antibody produced in rabbit, affinity isolated antibody
F8051 Anti-phospho-FAK (pTyr407) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4503913
Anti-phospho-FAK (pTyr407) antibody produced in rabbit, affinity isolated antibody
SAB4503869
Anti-phospho-FAK (pTyr576) antibody produced in rabbit, affinity isolated antibody
F8926 Anti-phospho-FAK (pTyr577) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
F9176 Anti-phospho-FAK (pTyr861) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
SAB4504404
Anti-phospho-FAK (pTyr861) antibody produced in rabbit, affinity isolated antibody
SAB4504405
Anti-phospho-FAK (pTyr925) antibody produced in rabbit, affinity isolated antibody
SAB4300281
Anti-phospho-PTK2 (pTyr576/pTyr577) antibody produced in rabbit, affinity isolated antibody
SAB4300047
Anti-phospho-PTK2 (pTyr861) antibody produced in rabbit, affinity isolated antibody
SAB4300101
Anti-phospho-PTK2 (pTyr925) antibody produced in rabbit, affinity isolated antibody
SAB4300433
Anti-SRC (Ab-529) antibody produced in rabbit, affinity isolated antibody
SAB2102295
Anti-SRC antibody produced in rabbit, affinity isolated antibody
SAB4502847
Anti-SRC antibody produced in rabbit, affinity isolated antibody
SAB2103351
Anti-SRC, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB1306786
ANTI-SRC1 (CENTER) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1300410
Anti-SRC1 (N-term) antibody produced in rabbit, saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution
SAB1306922
ANTI-SRC1(N-TERMINAL) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
F2006 Fibronectin from human plasma, lyophilized powder, BioReagent, suitable for cell culture
EMU028841 MISSION® esiRNA, esiRNA targeting mouse Rac1 (esiRNA1)
EHU075591 MISSION® esiRNA, esiRNA human RAC1 (esiRNA1)
SAB1402765
Monoclonal Anti-EPHB1 antibody produced in mouse, clone 4G6, purified immunoglobulin, buffered aqueous solution