EMAIL THIS PAGE TO A FRIEND
Journal of leukocyte biology
Δ⁹-Tetrahydrocannabinol-induced anti-inflammatory responses in adolescent mice switch to proinflammatory in adulthood.
PMID 24744434
Abstract
Marijuana abuse is prominent among adolescents. Although Δ(9)-THC, one of its main components, has been demonstrated to modulate immunity in adults, little is known about its impact during adolescence on the immune system and the long-lasting effects in adulthood. We demonstrate that 10 days of Δ(9)-THC treatment induced a similar alteration of macrophage and splenocyte cytokines in adolescent and adult mice. Immediately at the end of chronic Δ(9)-THC, a decrease of proinflammatory cytokines IL- 1β and TNF-α and an increase of anti-inflammatory cytokine IL-10 production by macrophages were present as protein and mRNA in adolescent and adult mice. In splenocytes, Δ(9)-THC modulated Th1/Th2 cytokines skewing toward Th2: IFN-γ was reduced, and IL-4 and IL-10 increased. These effects were lost in adult animals, 47 days after the last administration. In contrast, in adult animals treated as adolescents, a perturbation of immune responses, although in an opposite direction, was present. In adults treated as adolescents, a proinflammatory macrophage phenotype was observed (IL-1β and TNF-α were elevated; IL-10 decreased), and the production of Th cytokines was blunted. IgM titers were also reduced. Corticosterone concentrations indicate a long-lasting dysregulation of HPA in adolescent mice. We measured blood concentrations of Δ(9)-THC and its metabolites, showing that Δ(9)-THC plasma levels in our mice are in the order of those achieved in human heavy smokers. Our data demonstrate that Δ(9)-THC in adolescent mice triggers immune dysfunctions that last long after the end of abuse, switching the murine immune system to proinflammatory status in adulthood.