EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
The fungal chimerolectin MOA inhibits protein and DNA synthesis in NIH/3T3 cells and may induce BAX-mediated apoptosis.
PMID 24747075
Abstract
The Marasmius oreades mushroom agglutinin (MOA) is a blood group B-specific lectin carrying an active proteolytic domain. Its enzymatic activity has recently been shown to be critical for toxicity of MOA toward the fungivorous soil nematode Caenorhabditis elegans. Here we present evidence that MOA also induces cytotoxicity in a cellular model system (murine NIH/3T3 cells), by inhibiting protein synthesis, and that cytotoxicity correlates, at least in part, with proteolytic activity. A peptide-array screen identified the apoptosis mediator BAX as a potential proteolytic substrate and further suggests a variety of bacterial and fungal peptides as potential substrates. These findings are in line with the suggestion that MOA and related proteases may play a role for host defense.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E3132
E-64, protease inhibitor
C15H27N5O5