EMAIL THIS PAGE TO A FRIEND
Molecular immunology
Liver X receptor regulates mouse GM-CSF-derived dendritic cell differentiation in vitro.
PMID 24747959
Abstract
Liver X receptors (LXRs) are nuclear receptors that play an essential role in lipid and cholesterol metabolism. Emerging studies indicate a potential function for LXRs in regulating dendritic cell (DC)-dependent immune responses; however, the role of LXRs in DC differentiation is largely unknown. Here, we report that LXRα regulates the differentiation of mouse GM-CSF-derived DCs. Activation or overexpression of LXRα significantly enhanced myeloid DC differentiation from mouse bone marrow (BM) cells, while siRNA-mediated knockdown of LXRα suppressed DC differentiation. In addition, we demonstrated that LXR agonist-programmed DCs showed an increased capacity for stimulating T-cell proliferation. Mechanistic studies showed that activation of LXR could inhibit the phosphorylation of STAT3 and downregulate the expression of its target, S100A9, an important negative regulator of myeloid DC differentiation. We also found that Histone deacetylase (HDAC) inhibition interfered with the effect of LXR on STAT3 signaling via acetylation of STAT3. Chromatin immunoprecipitation assays confirmed that LXR activation and HDAC inhibition balanced the recruitment of STAT3 to the S100A9 promoter, which involved distinct post-translational modifications of STAT3. In conclusion, our observations demonstrated a novel role for LXRα in GM-CSF-derived DC differentiation and revealed the underlying mechanism.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C5041
6-Carboxyfluorescein diacetate, ~95% (HPLC)
C25H16O9
L5044
Anti-LXRα (N-terminal) antibody produced in rabbit, ~1.5 mg/mL, affinity isolated antibody, buffered aqueous solution
AV38906
Anti-NR1H2 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV45633
Anti-NR1H2 (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB1412663
ANTI-NR1H2 antibody produced in mouse, clone 1B6, purified immunoglobulin, buffered aqueous solution
SAB2103395
Anti-NR1H2 antibody produced in rabbit, affinity isolated antibody
HPA056838
Anti-NR1H2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1406923
Anti-NR1H3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV38670
Anti-NR1H3 antibody produced in rabbit, affinity isolated antibody
AV38669
Anti-NR1H3 antibody produced in rabbit, IgG fraction of antiserum
HPA036443
Anti-NR1H3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2106276
Anti-NR1H3 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
C4493
Cucurbitacin I hydrate, ≥95% (HPLC), solid
C30H42O7 · xH2O
EHU018341 MISSION® esiRNA, esiRNA targeting human NR1H2 (esiRNA1)
EMU008841 MISSION® esiRNA, esiRNA targeting mouse Nr1h2 (esiRNA1)
EHU134631 MISSION® esiRNA, esiRNA human NR1H3 (esiRNA1)
EMU073291 MISSION® esiRNA, esiRNA targeting mouse Nr1h3 (esiRNA1)
WH0007376M1
Monoclonal Anti-NR1H2 antibody produced in mouse, clone 2H2-H3, purified immunoglobulin, buffered aqueous solution
SAB1404516
Monoclonal Anti-NR1H2 antibody produced in mouse, clone 1E1, purified immunoglobulin, buffered aqueous solution
SAB1402495
Monoclonal Anti-NR1H3 antibody produced in mouse, clone 3E9, purified immunoglobulin, buffered aqueous solution