EMAIL THIS PAGE TO A FRIEND
Molecular immunology
Divergent roles of histone deacetylase 6 (HDAC6) and histone deacetylase 11 (HDAC11) on the transcriptional regulation of IL10 in antigen presenting cells.
PMID 24747960
Abstract
The anti-inflammatory cytokine IL-10 is a key modulator of immune responses. A better understanding of the regulation of this cytokine offers the possibility of tipping the balance of the immune response toward either tolerance, or enhanced immune responses. Histone deacetylases (HDACs) have been widely described as negative regulators of transcriptional regulation, and in this context, the primarily nuclear protein HDAC11 was shown to repress il-10 gene transcriptional activity in antigen-presenting cells (APCs). Here we report that another HDAC, HDAC6, primarily a cytoplasmic protein, associates with HDAC11 and modulates the expression of IL-10 as a transcriptional activator. To our knowledge, this is the first demonstration of two different HDACs being recruited to the same gene promoter to dictate divergent transcriptional responses. This dynamic interaction results in dynamic changes in the expression of IL-10 and might help to explain the intrinsic plasticity of the APC to determine T-cell activation versus T-cell tolerance.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06-599
Anti-acetyl-Histone H3 Antibody, 100 µg, Detect acetyl-Histone H3 with Anti-acetyl-Histone H3 Antibody (Rabbit Polyclonal Antibody), that has been shown to work in WB, ICC, ChIP, ChIP-seq.
SAB2502051
Anti-Hdac1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2701686
Anti-hdac1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701822
Anti-hdac1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AV38530
Anti-HDAC1 antibody produced in rabbit, IgG fraction of antiserum
SAB2101020
Anti-HDAC1 antibody produced in rabbit, affinity isolated antibody
HPA029693
Anti-HDAC1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1407989
Anti-HDAC11 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2101021
Anti-HDAC11 antibody produced in rabbit, affinity isolated antibody
SAB2104832
Anti-HDAC11 antibody produced in rabbit, affinity isolated antibody
SAB1406911
Anti-HDAC6 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV31451
Anti-HDAC6 antibody produced in rabbit, affinity isolated antibody
HPA026321
Anti-HDAC6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
HPA003714 Anti-HDAC6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
SAB2106255
Anti-HDAC6 antibody produced in rabbit, affinity isolated antibody
H3284
Anti-Histone Deacetylase 1 (HDAC1) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
H6287 Anti-Histone Deacetylase 1 (HDAC1) antibody, Mouse monoclonal, clone HDAC1-21, purified from hybridoma cell culture
H4539
Anti-Histone Deacetylase 11 (HDAC11) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
H2287
Anti-Histone Deacetylase 6 (HDAC6) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
F3290
FLAG® Peptide, lyophilized powder
C41H60N10O20
PLA0134
Rabbit anti-HDAC1 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.