EMAIL THIS PAGE TO A FRIEND
The Plant cell
The Enzyme-Like Domain of Arabidopsis Nuclear β-Amylases Is Critical for DNA Sequence Recognition and Transcriptional Activation.
PMID 24748042
Abstract
Plant BZR1-BAM transcription factors contain a β-amylase (BAM)-like domain, characteristic of proteins involved in starch breakdown. The enzyme-derived domains appear to be noncatalytic, but they determine the function of the two Arabidopsis thaliana BZR1-BAM isoforms (BAM7 and BAM8) during transcriptional initiation. Removal or swapping of the BAM domains demonstrates that the BAM7 BAM domain restricts DNA binding and transcriptional activation, while the BAM8 BAM domain allows both activities. Furthermore, we demonstrate that BAM7 and BAM8 interact on the protein level and cooperate during transcriptional regulation. Site-directed mutagenesis of residues in the BAM domain of BAM8 shows that its function as a transcriptional activator is independent of catalysis but requires an intact substrate binding site, suggesting it may bind a ligand. Microarray experiments with plants overexpressing truncated versions lacking the BAM domain indicate that the pseudo-enzymatic domain increases selectivity for the preferred cis-regulatory element BBRE (BZR1-BAM Responsive Element). Side specificity toward the G-box may allow crosstalk to other signaling networks. This work highlights the importance of the enzyme-derived domain of BZR1-BAMs, supporting their potential role as metabolic sensors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2500053
Anti-AKR1B10 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
AV42553
Anti-AKR1B10 antibody produced in rabbit, IgG fraction of antiserum
HPA020280
Anti-AKR1B10 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1411122
Anti-AKR1B10 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1405922
Anti-HAL antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV45694
Anti-HAL antibody produced in rabbit, IgG fraction of antiserum
SAB2101012
Anti-HAL antibody produced in rabbit, affinity isolated antibody
HPA038547
Anti-HAL antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA038548
Anti-HAL antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
WH0057016M1
Monoclonal Anti-AKR1B10 antibody produced in mouse, clone 1A6, purified immunoglobulin, buffered aqueous solution
SAB1405200
Monoclonal Anti-AKR1B10 antibody produced in mouse, clone 2B3, purified immunoglobulin, buffered aqueous solution
WH0003034M4
Monoclonal Anti-HAL antibody produced in mouse, clone 4F2, purified immunoglobulin, buffered aqueous solution