EMAIL THIS PAGE TO A FRIEND
Histochemistry and cell biology
Phosphatase and actin regulator 4 is associated with intermediate filaments in adult neural stem cells and their progenitor astrocytes.
PMID 24748504
Abstract
Phosphatase and actin regulator 4 (Phactr4) is a newly discovered protein that inhibits protein phosphatase 1 and shows actin-binding activity. We previously found that Phactr4 is expressed in the neurogenic niche in adult mice, although its precise subcellular localization and possible function in neural stem cells (NSCs) is not yet understood. Here, we show that Phactr4 formed punctiform clusters in the cytosol of subventricular zone-derived adult NSCs and their progeny in vitro. These Phactr4 signals were not associated with F-actin fibers but were closely associated with intermediate filaments such as nestin and glial fibrillary acidic protein (GFAP) fibers. Direct binding of Phactr4 with nestin and GFAP filaments was demonstrated using Duolink protein interaction analyses and immunoprecipitation assays. Interestingly, when nestin fibers were de-polymerized during the mitosis or by the phosphatase inhibitor, Phactr4 appeared to be dissociated from nestin, suggesting that their protein interaction is regulated by the protein phosphorylation. These results suggest that Phactr4 forms functional associations with intermediate filament networks in adult NSCs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
DUO92012
Duolink® In Situ Detection Reagents Brightfield
DUO92013
Duolink® In Situ Detection Reagents FarRed
DUO92014
Duolink® In Situ Detection Reagents Green
DUO92007
Duolink® In Situ Detection Reagents Orange
DUO92008
Duolink® In Situ Detection Reagents Red
DUO92106
Duolink® In Situ Orange Starter Kit Goat/Rabbit
DUO92104
Duolink® In Situ Orange Starter Kit Mouse/Goat
DUO92102
Duolink® In Situ Orange Starter Kit Mouse/Rabbit
DUO92006
Duolink® In Situ PLA® Probe Anti-Goat MINUS, Affinity purified Donkey anti-Goat IgG (H+L)
DUO92003
Duolink® In Situ PLA® Probe Anti-Goat PLUS, Affinity purified Donkey anti-Goat IgG (H+L)
DUO92004
Duolink® In Situ PLA® Probe Anti-Mouse MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
DUO92001
Duolink® In Situ PLA® Probe Anti-Mouse PLUS, Affinity purified Donkey anti-Mouse IgG (H+L)
DUO92005
Duolink® In Situ PLA® Probe Anti-Rabbit MINUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
DUO92002
Duolink® In Situ PLA® Probe Anti-Rabbit PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
DUO92010 Duolink® In Situ Probemaker MINUS
DUO92009 Duolink® In Situ Probemaker PLUS
DUO92105
Duolink® In Situ Red Starter Kit Goat/Rabbit
DUO92103
Duolink® In Situ Red Starter Kit Mouse/Goat
DUO92101
Duolink® In Situ Red Starter Kit Mouse/Rabbit