EMAIL THIS PAGE TO A FRIEND
Parasite immunology
A multivalent chimeric vaccine composed of Schistosoma mansoni SmTSP-2 and Sm29 was able to induce protection against infection in mice.
PMID 24749785
Abstract
Schistosoma mansoni is a blood fluke parasite responsible for schistosomiasis. The best long-term strategy to control schistosomiasis is through immunization combined with drug treatment. In this study, we cloned, expressed and purified SmTSP-2 fused to the N- and C-terminal halves of Sm29 and tested these chimeras as vaccine candidates using an adjuvant approved to be used in humans. The results demonstrated that vaccination with SmTSP-2 fused to N- or C-terminus of Sm29-induced reduction in worm burden and liver pathology when compared to control animals. Additionally, we detected high levels of mouse-specific IgG, IgG1 and IgG2a against both chimeras and significant amounts of IFN-γ and TNF-α and no IL-4. Finally, studies with sera from patients resistant to infection and living in schistosomiasis endemic areas revealed high levels of specific IgG to both chimeras when compared to healthy individuals. In conclusion, SmTSP-2/Sm29 chimeras tested here induced partial protection against infection and might be a potential vaccine candidate.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A3682 Anti-Mouse IgG (Fab specific)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3701029 Anti-Mouse IgG (Fc specific), F(ab′)2 fragment, highly cross adsorbed-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
A9309 Anti-Mouse IgG (Fc specific)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3701023 Anti-Mouse IgG (Fc specific)-Peroxidase antibody produced in rabbit, affinity isolated antibody, lyophilized powder
A3673 Anti-Mouse IgG (γ-chain specific)−Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3701037 Anti-Mouse IgG (γ-chain specific)-Peroxidase antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3701131 Anti-Mouse IgG (H+L), F(ab′)2 fragment-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3701159 Anti-Mouse IgG (H+L), F(ab) fragment-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3701165 Anti-Mouse IgG (H+L), F(ab) fragment-Peroxidase antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3701116 Anti-Mouse IgG (H+L)-Peroxidase antibody produced in chicken, affinity isolated antibody, lyophilized powder
SAB3701105
Anti-Mouse IgG (H+L)-Peroxidase antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB3701083
Anti-Mouse IgG (H+L)-Peroxidase antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3701095
Anti-Mouse IgG (H+L)-Peroxidase antibody produced in sheep, affinity isolated antibody, lyophilized powder
A8924 Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
A5278 Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution
A5906 Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in sheep, affinity isolated antibody, buffered aqueous solution
A6782 Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in sheep, affinity isolated antibody, buffered aqueous solution
SAB3701015 Anti-Mouse IgG F(ab′)2, F(ab′)2 fragment, highly cross absorbed-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3701004 Anti-Mouse IgG F(ab′)2, highly cross adsorbed-Peroxidase antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB3700993 Anti-Mouse IgG F(ab′)2-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3701003 Anti-Mouse IgG F(ab′)2-Peroxidase antibody produced in rabbit, affinity isolated antibody, lyophilized powder
78410
o-Phenylenediamine, suitable for fluorescence, ≥99.0% (NT)
C6H8N2