EMAIL THIS PAGE TO A FRIEND
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Expression of Lgr5, a marker of intestinal stem cells, in colorectal cancer and its clinicopathological significance.
PMID 24751002
Abstract
Cancer stem cells (CSCs) have been the focus of intense investigations in cancer research although the cellular origin of CSCs has not been clearly determined. Lgr5 is a regulated target of Wnt/β-catenin signaling, which was first identified as a marker of intestinal stem cells. However, the expression of Lgr5 in human colorectal cancer (CRC) and its clinical clinicopathological significance in CRC patients as well as its correlation with Wnt/β-catenin pathway are not fully explored. Localization and expression of Lgr5 in CRC tissues was performed by immunohistochemical staining. The correlation between its expression levels and clinicopathological features as well as clinical outcomes of patients was analysed. The quantitative expression levels of Lgr5 in various CRC cell lines were determined using real-time RT-PCR. The relationship between Lgr5 expression and Wnt/β-catenin pathway in CRC was also investigated. Obviously elevated expression of Lgr5 was observed in CRC tissues, compared to the paired nontumor tissues. mRNA expression levels of Lgr5 was positively correlated with the expression of β-catenin in CRC tissues. The expression of Lgr5 was various in different CRC cell lines. Low and high expression levels of Lgr5 were significantly correlated with clinicopathological features such as TNM stage, lymph node metastasis and vascular invasion of CRC patients. More importantly, Lgr5 expression in CRC tissues was also associated with tumor angiogenesis as well as clinical outcomes. Taken together, these results suggest that elevated Lgr5 expression might contribute to the development and progression of CRC, and it could also be used a potential unfavorable prognostic biomarker for CRC. A better understanding of molecule mechanisms and the relevance of potential value for Lgr5 in the progression of CRC will help to identify a novel therapeutic strategy for CRC patients.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB3700857 Anti-Rabbit IgG (Fc specific), highly cross adsorbed-Biotin antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700856 Anti-Rabbit IgG (Fc specific)-Biotin antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB4600006 Anti-Rabbit IgG (H+L), Biotin antibody produced in goat, ~2 mg/mL, affinity isolated antibody
SAB3700951 Anti-Rabbit IgG (H+L), F(ab′)2 fragment-Biotin antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700973 Anti-Rabbit IgG (H+L), F(ab) fragment-Biotin antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB4600007 Anti-Rabbit IgG (H+L), highly cross-adsorbed, Biotin antibody produced in goat, ~2 mg/mL, affinity isolated antibody
B7389 Anti-Rabbit IgG (whole molecule)–Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
B8895 Anti-Rabbit IgG (whole molecule)–Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB3700844 Anti-Rabbit IgG F(ab′)2, F(ab′)2 fragment, highly cross absorbed-Biotin antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700836 Anti-Rabbit IgG F(ab′)2, highly cross adsorbed-Biotin antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700835 Anti-Rabbit IgG F(ab′)2-Biotin antibody produced in goat, affinity isolated antibody, lyophilized powder