EMAIL THIS PAGE TO A FRIEND
Vascular pharmacology
Tirofiban induces VEGF production and stimulates migration and proliferation of endothelial cells.
PMID 24751361
Abstract
Tirofiban is a fibrinogen receptor antagonist, generated using the tripeptide Arg-Gly-Asp (RGD) as a template. RGD activates integrin receptors and integrin-mediated signals are necessary for normal cells to promote survival and stimulate cell cycle progression. We investigated whether tirofiban activated growth-stimulatory signals in endothelium. For this study human umbilical vein endothelial cells (HUVEC) and human aortic endothelial cells (HAEC) were used. Analysis of cell proliferation, by cell counts, showed that the number of endothelial cells doubled after 72 h of culture in the absence of tirofiban, while they were tripled and even quadrupled, in the presence of increasing doses of the drug. Moreover, tirofiban-stimulated cells had a greater ability to migrate through the transwell filters of Boyden chamber, compared to unstimulated cells. The scratch assay, which mimics cell migration during wound healing, showed that tirofiban stimulated HUVECs to migrate into the leading hedge of the scratch. Western blot showed that tirofiban increased the expression levels of VEGF and the downstream effectors Erk and cyclin D. An inhibitor of VEGFR2 counteracted tirofiban-induced-proliferation, suggesting a role for VEGF in such effect. Our study shows that tirofiban stimulates the migration and proliferation of endothelial cells suggesting that it can promote endothelial repair. Ex vivo cultures of arterial rings confirmed the growth stimulatory effect of tirofiban on endothelium. Thus, the benefits of tirofiban in those with acute coronary syndromes undergoing PTCA may be due to rapid endothelization of damaged vessel, besides antiplatelet effects.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB3700853 Anti-Rabbit IgG (Fc specific), highly cross adsorbed-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700863 Anti-Rabbit IgG (Fc specific)-Peroxidase antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB3700852 Anti-Rabbit IgG (Fc specific)-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700949 Anti-Rabbit IgG (H+L), F(ab′)2 fragment-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700978 Anti-Rabbit IgG (H+L), F(ab) fragment-Peroxidase antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB3700972 Anti-Rabbit IgG (H+L), F(ab) fragment-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700941 Anti-Rabbit IgG (H+L)-Peroxidase antibody produced in chicken, affinity isolated antibody, lyophilized powder
SAB3700934 Anti-Rabbit IgG (H+L)-Peroxidase antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB3700920 Anti-Rabbit IgG (H+L)-Peroxidase antibody produced in sheep, affinity isolated antibody, lyophilized powder
A6667 Anti-Rabbit IgG (whole molecule), F(ab′)2 fragment−Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
A4914 Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
A8275 Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution
SAB3700842
Anti-Rabbit IgG F(ab′)2, F(ab′)2 fragment, highly cross absorbed-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700832 Anti-Rabbit IgG F(ab′)2, highly cross adsorbed-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700831 Anti-Rabbit IgG F(ab′)2-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
V4512 Vascular Endothelial Growth Factor from mouse, VEGF, recombinant, expressed in E. coli, powder, suitable for cell culture
V7259 Vascular Endothelial Growth Factor human, VEGF, recombinant, expressed in E. coli, powder, suitable for cell culture
SRP3182
VEGF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), cell culture tested