EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
The number of responding CD4 T cells and the dose of antigen conjointly determine the TH1/TH2 phenotype by modulating B7/CD28 interactions.
PMID 24752446
Abstract
Our previous in vivo studies show that both the amount of Ag and the number of available naive CD4 T cells affect the Th1/Th2 phenotype of the effector CD4 T cells generated. We examined how the number of OVA-specific CD4 TCR transgenic T cells affects the Th1/Th2 phenotype of anti-SRBC CD4 T cells generated in vivo upon immunization with different amounts of OVA-SRBC. Our observations show that a greater number of Ag-dependent CD4 T cell interactions are required to generate Th2 than Th1 cells. We established an in vitro system that recapitulates our main in vivo findings to more readily analyze the underlying mechanism. The in vitro generation of Th2 cells depends, as in vivo, upon both the number of responding CD4 T cells and the amount of Ag. We demonstrate, using agonostic/antagonistic Abs to various costimulatory molecules or their receptors, that the greater number of CD4 T cell interactions, required to generate Th2 over Th1 cells, does not involve CD40, OX40, or ICOS costimulation, but does involve B7/CD28 interactions. A comparison of the level of expression of B7 molecules by APC and CD4 T cells, under different conditions resulting in the substantial generation of Th1 and Th2 cells, leads us to propose that the critical CD28/B7 interactions, required to generate Th2 cells, may directly occur between CD4 T cells engaged with the same B cell acting as an APC.