EMAIL THIS PAGE TO A FRIEND
Cutaneous and ocular toxicology
Inhibitory effects of the platelet-activating factor receptor antagonists, CV-3988 and Ginkgolide B, on alkali burn-induced corneal neovascularization.
PMID 24754407
Abstract
Platelet-activating factor (PAF) has been found in various ocular tissues; the activity of PAF depends on the binding to its specific receptor, PAF-receptor. We investigated the therapeutic effects of PAF-receptor antagonists (CV-3988 and Ginkgolide B) on alkali burn-induced corneal neovascularization (CNV). CNV was induced by applying a 0.2 N sodium hydroxide (3 µl, NaOH) solution directly on mice corneas. CV-3988 (1 mM/10 µl) and Ginkgolide B (1 mM/10 µl) were administered topically on the corneas three times daily for three consecutive days. CNV was evaluated under a slit-lamp microscope. Corneas were processed for histological, immunohistochemical and reverse transcription polymerase chain reaction analysis. Human umbilical vein endothelial cells were used for the migration and tube formation assay. Application of CV-3988 and Ginkgolide B inhibited CNV caused by alkali burn. CV-3988 and Ginkgolide B attenuated the expression of PAF-receptor mRNA. Alkali injury induced a massively increased intraocular mRNA expression of an angiogenic factor in cornea tissues, whereas these increments were attenuated by the application of CV-3988 and Ginkgolide B. CV-3988 and Ginkgolide B reversed opacity and neovascularization in alkali burn-induced corneas. Our findings suggest that CV-3988 and Ginkgolide B may be therapeutically useful in the treatment of CNV and inflammation.