EMAIL THIS PAGE TO A FRIEND
European journal of clinical investigation
C3 and C4 are strongly related to adipose tissue variables and cardiovascular risk factors.
PMID 24754458
Abstract
In several reports, C3 and C4 have been linked to diabetes and cardiovascular disease (CVD). Here, we investigate this link and the degree of C3 activation in elderly individuals. In this study, C3 and C4 and the activation fragment C3a-desArg were analysed in 1016 subjects aged 70, in which blood pressure, lipid variables and fasting blood glucose were assessed. C3 levels were related to all the investigated classical cardiovascular risk factors and the metabolic syndrome (BMI, waist circumference, fat distribution, blood pressure, blood glucose levels, TG) except total cholesterol and LDL cholesterol in a highly significant fashion (Spearman up to 0,5; Pxa0