EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Differentiation, polarization, and migration of human induced pluripotent stem cell-derived neural progenitor cells co-cultured with a human glial cell line with radial glial-like characteristics.
PMID 24755070
Abstract
Here we established a unique human glial cell line, GDC90, derived from a human glioma and demonstrated its utility as a glial scaffold for the polarization and differentiation of human induced pluripotent stem cell-derived neural progenitor cells (iPSC-NPCs). When co-cultured with GDC90 cells, iPSC-NPCs underwent rapid polarization and neurite extension along the radially spreading processes of the GDC90 cells, and showed migratory behavior. This method is potentially useful for detailed examination of neurites or for controlling neurites behavior for regenerative medicine.