EMAIL THIS PAGE TO A FRIEND
International journal of medical microbiology : IJMM
Dynamin-related protein Drp1 and mitochondria are important for Shigella flexneri infection.
PMID 24755420
Abstract
Shigella infection in epithelial cells induces cell death which is accompanied by mitochondrial dysfunction. In this study the role of the mitochondrial fission protein, Drp1 during Shigella infection in HeLa cells was examined. Significant lactate dehydrogenase (LDH) release was detected in the culture supernatant when HeLa cells were infected with Shigella at a high multiplicity of infection. Drp1 inhibition with Mdivi-1 and siRNA knockdown significantly reduced LDH release. HeLa cell death was also accompanied by mitochondrial fragmentation. Tubular mitochondrial networks were partially restored when Drp1 was depleted with either siRNA or inhibited with Mdivi-1. Surprisingly either Mdivi-1 treatment or Drp1 siRNA-depletion of HeLa cells also reduced Shigella plaque formation. The effect of Mdivi-1 on Shigella infection was assessed using the murine Sereny model, however it had no impact on ocular inflammation. Overall our results suggest that Drp1 and the mitochondria play important roles during Shigella infection.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1400649
Anti-C6orf210 antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
HPA039324
Anti-DNM1L antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB3501074
Anti-DNM1L antibody produced in rabbit, affinity isolated antibody
SAB2105834
Anti-DNM1L, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB2101766
Anti-PDSS2 antibody produced in rabbit, affinity isolated antibody
HPA029685
Anti-PDSS2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1
HPA029686
Anti-PDSS2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab2
SML0412
IM-54, ≥98% (HPLC)
C19H23N3O2
M0199
Mdivi-1, ≥98% (HPLC), powder
C15H10Cl2N2O2S
EHU081751 MISSION® esiRNA, esiRNA human DNM1L (esiRNA1)
EMU084221 MISSION® esiRNA, esiRNA targeting mouse Dnm1l (esiRNA1)
WH0010059M1
Monoclonal Anti-DNM1L antibody produced in mouse, clone 3B5, purified immunoglobulin, buffered aqueous solution
SAB1404777
Monoclonal Anti-DNM1L antibody produced in mouse, clone 1D6, purified immunoglobulin, buffered aqueous solution
SAB4100593
Monoclonal Anti-DNM1L antibody produced in mouse, clone DNM1L-A573, culture supernatant
N9037
Necrostatin-1, ≥98% (HPLC)
C13H13N3OS
78429
p-Phenylenediamine, ≥99.0% (GC/NT)
C6H8N2
P6001
p-Phenylenediamine
C6H8N2
78430
p-Phenylenediamine, technical, ≥97.0% (GC/NT)
C6H8N2
695106
p-Phenylenediamine, sublimed, ≥99%
C6H8N2