EMAIL THIS PAGE TO A FRIEND
Acta crystallographica. Section E, Structure reports online
4-Cyano-3-fluoro-phenyl 4-(hexa-dec-yl-oxy)benzoate.
PMID 24764965
Abstract
In the title compound, C30H40FNO3, the dihedral angle between the benzene rings is 57.76 (7)°. The alkyl chain adopts an all-trans conformation. In the crystal, mol-ecules are linked by pairs of C-H⋯O hydrogen bonds, forming inversion dimers.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
417440
2-Fluoro-4-hydroxybenzonitrile, 99%
C7H4FNO