EMAIL THIS PAGE TO A FRIEND
The Journal of allergy and clinical immunology
CCR10 regulates balanced maintenance and function of resident regulatory and effector T cells to promote immune homeostasis in the skin.
PMID 24767879
Abstract
CCR10 and CCL27 make up the most skin-specific chemokine receptor/ligand pair implicated in skin allergy and inflammatory diseases, including atopic dermatitis and psoriasis. This pair is thought to regulate the migration, maintenance, or both of skin T cells and is suggested to be therapeutic targets for treatment of skin diseases. However, the functional importance of CCR10/CCL27 in vivo remains elusive. We sought to determine the expression and function of CCR10 in different subsets of skin T cells under both homeostatic and inflammatory conditions to gain a mechanistic insight into the potential roles of CCR10 during skin inflammation. Using heterozygous and homozygous CCR10 knockout/enhanced green fluorescent protein knockin mice, we assessed the expression of CCR10 on regulatory and effector T cells of healthy and inflamed skin induced by chemicals, pathogens, and autoreactive T cells. In addition, we assessed the effect of CCR10 knockout on the maintenance and functions of different T cells and inflammatory status in the skin during different phases of the immune response. CCR10 expression is preferentially induced on memory-like skin-resident T cells and their progenitors for their maintenance in homeostatic skin but not expressed on most skin-infiltrating effector T cells during inflammation. In CCR10 knockout mice the imbalanced presence and dysregulated function of resident regulatory and effector T cells result in over-reactive and prolonged innate and memory responses in the skin, leading to increased clearance of Leishmania species infection in the skin. CCR10 is a critical regulator of skin immune homeostasis.