EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Polymeric ionic liquid bucky gels as sorbent coatings for solid-phase microextraction.
PMID 24768123
Abstract
Novel cross-linked polymeric ionic liquid (PIL) bucky gels were formed by free-radical polymerization of polymerizable ionic liquids gelled with multi-walled carbon nanotubes (MWCNT) and used as sorbent coatings for solid-phase microextraction (SPME). The combination of PIL with MWCNTs significantly enhanced the π-π interaction between the sorbent coatings and the aromatic analytes. Compared to the neat PIL-based sorbent coating, the PIL bucky gel sorbent coatings demonstrated higher extraction efficiency for the extraction of polycyclic aromatic hydrocarbons (PAHs). A partitioning extraction mechanism was observed for the PIL/MWCNT-based sorbent coatings indicating that the addition of MWCNTs did not seem to affect the extraction mechanism of the sorbent coating. The analyte-to-coating partition coefficients (logKfs) were estimated and the limits of detection (LOD) for selected PIL bucky gel sorbent coating were determined to be in the range of 1-2.5 ng L(-1). Recovery studies were also performed for PAHs in river and tap water to validate the applicability of the developed method.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
08953
1-Bromobutane, analytical standard
C4H9Br
B59497
1-Bromobutane, ReagentPlus®, 99%
C4H9Br
95446
1-Octanol, analytical standard
C8H18O
74850
1-Octanol, puriss., ≥99.5% (GC)
C8H18O
297887
1-Octanol, anhydrous, ≥99%
C8H18O
W280003
1-Octanol, ≥98%, FCC, FG
C8H18O
W280011
1-Octanol, natural, ≥98%, FCC
C8H18O
235466
1-Vinylimidazole, ≥99%
C5H6N2
215376
Acenaphthene, 99%
C12H10
05426
Acenaphthene, certified reference material, TraceCERT®
C12H10
48500-U
Acenaphthene, analytical standard
C12H10
455830
Ammonium hydrogen difluoride, 99.999% trace metals basis
H5F2N
224820
Ammonium hydrogen difluoride, reagent grade, 95%
H5F2N
01110
Ammonium hydrogen difluoride, puriss., ≥97.5%
H5F2N
07671
Anthracene, certified reference material, TraceCERT®
C14H10
31581
Anthracene, analytical standard
C14H10
48567
Anthracene, analytical standard
C14H10
10580
Anthracene, suitable for scintillation, ≥99.0% (GC)
C14H10
331481
Anthracene, zone-refined, ≥99%
C14H10
141062
Anthracene, ReagentPlus®, 99%
C14H10
A89200
Anthracene, reagent grade, 97%
C14H10
694959
Anthracene, sublimed grade, ≥99%
C14H10
ERA-010
Anthracene, vial of 250 mg, analytical standard
C14H10
40076
Anthracene solution, certified reference material, 1000 μg/mL in acetone
C14H10
48647
Anthracene solution, certified reference material, 200 μg/mL in methanol
C14H10
35754
Chrysene, analytical standard
C18H12
245186
Chrysene, 98%
C18H12
48565-U
Chrysene, analytical standard
C18H12
BCR269
Chrysene, BCR® certified Reference Material
C18H12
94035
Chrysene, certified reference material, TraceCERT®
C18H12