EMAIL THIS PAGE TO A FRIEND
Translational research : the journal of laboratory and clinical medicine
RNA epigenetics.
PMID 24768686
Abstract
Mammalian messenger RNA (mRNA) and long noncoding RNA (lncRNA) contain tens of thousands of posttranscriptional chemical modifications. Among these, the N(6)-methyl-adenosine (m(6)A) modification is the most abundant and can be removed by specific mammalian enzymes. m(6)A modification is recognized by families of RNAxa0binding proteins that affect many aspects of mRNA function. mRNA/lncRNA modification represents another layer of epigenetic regulation of gene expression, analogous to DNA methylation and histone modification.