EMAIL THIS PAGE TO A FRIEND
Water research
Comparison of a novel extraction-based colorimetric (ABTS) method with membrane introduction mass spectrometry (MIMS): trichloramine dynamics in pool water.
PMID 24769062
Abstract
Trichloramine is a hazardous disinfection by-product, which is present in chlorinated swimming pools. Although it is primarily taken up by inhalation, the concentration in pool water is important to monitor pool water quality and to assess trichloramine mitigation strategies. To date, scarce data is available on trichloramine concentration in pool water due to the lack of a suitable and easily applicable analytical method. This study presents a novel low cost, colorimetric method which is easy to operate and suitable for on-site measurements of trichloramine concentrations ≥0.05xa0μM (≥0.01xa0mgxa0L(-1) as Cl2). The analytical method (termed "extraction-based ABTS method") consists of, (i) trichloramine stripping from pool water samples, (ii) passing it through a solid phase filter, composed of silica gel impregnated with sulfamic acid to eliminate interferences and (iii) trichloramine reaction with the indicator 2,2-azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt (ABTS) to produce the coloured ABTS(•-) radical, which is measured at λxa0=xa0405xa0nm to determine the trichloramine concentration in the pool water sample. A comparison of the extraction-based ABTS method with membrane introduction mass spectrometry (MIMS) for 28 pool samples revealed a good correlation of the two methods. The trichloramine concentration in pool samples is correlated to HOCl, which is the most important factor for its formation. Other parameters such as combined chlorine and pH play a minor role while no correlation between trichloramine and the urea or the TOC concentration was observed. On-site measurements with MIMS in a wading pool over 6 days with a time resolution of 1xa0h confirmed that trichloramine concentrations strongly responded to changes in free chlorine concentrations. A diurnal measurement of trichloramine with a time resolution of 20xa0min revealed that trichloramine concentrations reacted quickly and sensitively to the bather load and that urea is probably not the main precursor for its formation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
09702
Ammonium chloride, purum p.a., ≥99.0%
H4ClN
PHR1327
Ammonium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
H4ClN
09724
Ammonium chloride, tested according to Ph.Eur.
H4ClN
09683
Ammonium ion solution for ISE, 0.1 M NH4+, analytical standard (for ion-selective electrodes)
H4ClN
09685
Ammonium ion solution for ISE, 1000 mg/kg N, analytical standard (for ion-selective electrodes)
H4ClN
22754
Chlorine, puriss., ≥99.8%
Cl2
295132
Chlorine, ≥99.5%
Cl2
311421
Perchloric acid, 70%, 99.999% trace metals basis
HClO4
50439
Perchloric acid concentrate, 0.01 M HClO4 in water (0.01N), eluent concentrate for IC
HClO4
03366
Phosphate atomic spectroscopy standard concentrate 1.00 g PO43-, 1.00 g/L, for 1L standard solution, analytical standard
H3O4P
P5811
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
H3O4P
466123
Phosphoric acid, crystalline, ≥99.999% trace metals basis
H3O4P
452289
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
H3O4P
345245
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
H3O4P
79617
Phosphoric acid, BioUltra, ≥85% (T)
H3O4P
04102
Phosphoric acid, puriss., ≥99%
H3O4P
40278
Phosphoric acid, semiconductor grade MOS PURANAL (Honeywell 17938), ≥85%
H3O4P
40266
Phosphoric acid, semiconductor grade PURANAL (Honeywell 17861), ≥85%
H3O4P
612162
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
H3O4P
699608
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
H3O4P
698822
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
H3O4P
W290017
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
H3O4P
609781
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
H316O4P
RDD007
Sodium phosphate monobasic, anhydrous, free-flowing, Redi-Dri, ≥99.0%
H2NaO4P
S8282
Sodium phosphate monobasic, BioXtra, ≥99.0%
H2NaO4P
S3139
Sodium phosphate monobasic, BioReagent, for molecular biology, anhydrous, ≥98%
H2NaO4P
S5011
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
H2NaO4P
S2554
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
H2NaO4P
S6566
Sodium phosphate monobasic, Biotechnology Performance Certified, Cell Culture Tested
H2NaO4P
229903
Sodium phosphate monobasic, 99.999% trace metals basis
H2NaO4P