EMAIL THIS PAGE TO A FRIEND
The Journal of pathology
Mixed-species biofilm compromises wound healing by disrupting epidermal barrier function.
PMID 24771509
Abstract
In chronic wounds, biofilm infects host tissue for extended periods of time. This work establishes the first chronic preclinical model of wound biofilm infection aimed at addressing the long-term host response. Although biofilm-infected wounds did not show marked differences in wound closure, the repaired skin demonstrated compromised barrier function. This observation is clinically significant, because it leads to the notion that even if a biofilm infected wound is closed, as observed visually, it may be complicated by the presence of failed skin, which is likely to be infected and/or further complicated postclosure. Study of the underlying mechanisms recognized for the first time biofilm-inducible miR-146a and miR-106b in the host skin wound-edge tissue. These miRs silenced ZO-1 and ZO-2 to compromise tight junction function, resulting in leaky skin as measured by transepidermal water loss (TEWL). Intervention strategies aimed at inhibiting biofilm-inducible miRNAs may be productive in restoring the barrier function of host skin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
29238
Activated Charcoal Norit®, Norit RBAA-3, rod
C
GF93740384
Carbon - Vitreous, rod, 100mm, diameter 10mm, glassy carbon
C
GF65427587
Carbon - Vitreous, rod, 100mm, diameter 3.0mm, glassy carbon
C
GF05962765
Carbon - Vitreous, rod, 100mm, diameter 5.0mm, glassy carbon
C
GF68195403
Carbon - Vitreous, rod, 100mm, diameter 7.0mm, glassy carbon
C
GF23723423
Carbon - Vitreous, rod, 200mm, diameter 1.0mm, glassy carbon
C
GF91785134
Carbon - Vitreous, rod, 200mm, diameter 10mm, glassy carbon
C
GF78447707
Carbon - Vitreous, rod, 200mm, diameter 3.0mm, glassy carbon
C
GF45345562
Carbon - Vitreous, rod, 200mm, diameter 5.0mm, glassy carbon
C
GF24425805
Carbon - Vitreous, rod, 200mm, diameter 7.0mm, glassy carbon
C
GF93251011
Carbon - Vitreous, rod, 50mm, diameter 1.0mm, glassy carbon
C
GF20176680
Carbon - Vitreous, rod, 5mm, diameter 3.0mm, glassy carbon
C
GF59400233
Carbon - Vitreous, tube, 100mm, outside diameter 10mm, inside diameter 3mm, wall thickness 3.5mm, glassy carbon
C
GF39088648
Carbon - Vitreous, tube, 50mm, outside diameter 10mm, inside diameter 3mm, wall thickness 3.5mm, glassy carbon
C
GF07378476
Carbon - Vitreous, foam, 150x150mm, thickness 2.5mm, bulk density 0.05g/cm3, porosity 96.5%
C
GF39873369
Carbon - Vitreous, foam, 150x150mm, thickness 3.2mm, bulk density 0.05g/cm3, porosity 96.5%
C
GF61342220
Carbon - Vitreous, foam, 150x150mm, 0.05g.cmué, porosity 96.5%, 24 pores/cm
C
GF58042324
Carbon - Vitreous, foam, 275x330mm, thickness 3.2mm, bulk density 0.05g/cm3, porosity 96.5%
C
GF12408713
Carbon - Vitreous, foam, 275x330mm, 0.05g.cmué, porosity 96.5%, 24 pores/cm
C
GF04031132
Carbon - Vitreous, foam, 300x300mm, thickness 20mm, bulk density 0.05g/cm3, porosity 96.5%
C
GF06230922
Carbon - Vitreous, foam, 300x300mm, thickness 30mm, bulk density 0.05g/cm3, porosity 96.5%
C
GF95357526
Carbon - Vitreous, foil, 100x100mm, thickness 1.0mm, glassy carbon
C
GF61808889
Carbon - Vitreous, foil, 100x100mm, thickness 2.0mm, glassy carbon
C
GF09325009
Carbon - Vitreous, foil, 100x100mm, thickness 6.0mm, glassy carbon
C
GF22691426
Carbon - Vitreous, foil, 10x10mm, thickness 1.0mm, glassy carbon
C
GF74135628
Carbon - Vitreous, foil, 10x10mm, thickness 2.0mm, glassy carbon
C
GF99431523
Carbon - Vitreous, foil, 10x10mm, thickness 4.0mm, glassy carbon
C
GF44361457
Carbon - Vitreous, foil, 200x200mm, thickness 1.0mm, glassy carbon
C
GF10355958
Carbon - Vitreous, foil, 25x25mm, thickness 0.5mm, glassy carbon
C
GF89520521
Carbon - Vitreous, foil, 25x25mm, thickness 4.0mm, glassy carbon
C