EMAIL THIS PAGE TO A FRIEND
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Ferroportin mediates the intestinal absorption of iron from a nanoparticulate ferritin core mimetic in mice.
PMID 24776745
Abstract
The ferritin core is composed of fine nanoparticulate Fe(3+) oxohydroxide, and we have developed a synthetic mimetic, nanoparticulate Fe(3+) polyoxohydroxide (nanoFe(3+)). The aim of this study was to determine how dietary iron derived in this fashion is absorbed in the duodenum. Following a 4 wk run-in on an Fe-deficient diet, mice with intestinal-specific disruption of the Fpn-1 gene (Fpn-KO), or littermate wild-type (WT) controls, were supplemented with Fe(2+) sulfate (FeSO4), nanoFe(3+), or no added Fe for a further 4 wk. A control group was Fe sufficient throughout. Direct intestinal absorption of nanoFe(3+) was investigated using isolated duodenal loops. Our data show that FeSO4 and nanoFe(3+) are equally bioavailable in WT mice, and at wk 8 the mean ± SEM hemoglobin increase was 18 ± 7 g/L in the FeSO4 group and 30 ± 5 g/L in the nanoFe(3+) group. Oral iron failed to be utilized by Fpn-KO mice and was retained in enterocytes, irrespective of the iron source. In summary, although nanoFe(3+) is taken up directly by the duodenum its homeostasis is under the normal regulatory control of dietary iron absorption, namely via ferroportin-dependent efflux from enterocytes, and thus offers potential as a novel oral iron supplement.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
09582
Adipic acid, BioXtra, ≥99.5% (HPLC)
C6H10O4
89143
Adipic acid, certified reference material, TraceCERT®
C6H10O4
Y0000110
Adipic acid, European Pharmacopoeia (EP) Reference Standard
C6H10O4
A26357
Adipic acid, 99%
C6H10O4
W201103
Adipic acid, ≥99.5%
C6H10O4
F3388
Ferric citrate, BioReagent, suitable for cell culture
C6H5FeO7
RES4055F-A7
Ferric citrate, PharmaGrade, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.
C6H5FeO7 · H2O
121398
Giemsa stain, certified by the Biological Stain Commission
C14H14ClN3S
G4507
Giemsa stain, technical grade, used as a blood stain
C14H14ClN3S
G9641
Giemsa stain, powder, BioReagent, suitable for cell culture
C14H14ClN3S
G5637
Giemsa stain, certified by the Biological Stain Commission
C14H14ClN3S
11700
Giemsa stain, for microscopy (Bact., Hist.)
C14H14ClN3S
F8263
Iron(II) sulfate heptahydrate, plant cell culture tested, ≥99%
FeO4S · 7H2O
F8633
Iron(II) sulfate heptahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
FeO4S · 7H2O
44970
Iron(II) sulfate heptahydrate, puriss. p.a., ACS reagent
FeO4S · 7H2O
215422
Iron(II) sulfate heptahydrate, ACS reagent, ≥99.0%
FeO4S · 7H2O
F7002
Iron(II) sulfate heptahydrate, ReagentPlus®, ≥99%
FeO4S · 7H2O
31236
Iron(II) sulfate heptahydrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.0-103.4% (manganometric)
FeO4S · 7H2O
12354
Iron(II) sulfate heptahydrate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.5-104.5% (manganometric)
FeO4S · 7H2O
F6129
Iron(III) citrate, technical grade
C6H5FeO7
44941
Iron(III) citrate tribasic monohydrate, 18-20% Fe basis (T)
C6H5FeO7 · H2O