EMAIL THIS PAGE TO A FRIEND
European journal of immunology
Static and dynamic components synergize to form a stable survival niche for bone marrow plasma cells.
PMID 24777940
Abstract
In the bone marrow (BM), memory plasma cells (PCs) survive for long time periods in dedicated microenvironmental survival niches, resting in terms of proliferation. Several cell types, such as eosinophils and reticular stromal cells, have been reported to contribute to the survival niche of memory PCs. However, until now it has not been demonstrated whether the niche is formed by a fixed cellular microenvironment. By intravital microscopy, we provide for the first time evidence that the direct contacts formed between PCs and reticular stromal cells are stable in vivo, and thus the PCs are sessile in their niches. The majority (∼ 80%) of PCs directly contact reticular stromal cells in a non-random fashion. The mesenchymal reticular stromal cells in contact with memory PCs are not proliferating. On the other hand, we show here that eosinophils in the vicinity of long-lived PCs are vigorously proliferating cells and represent a dynamic component of the survival niche. In contrast, if eosinophils are depleted by irradiation, newly generated eosinophils localize in the vicinity of radiation-resistant PCs and the stromal cells. These results suggest that memory PC niches may provide attraction for eosinophils to maintain stability with fluctuating yet essential accessory cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1401144
Anti-FPR2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA029154 Anti-FPR2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV43141
Anti-MKRN1 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV43142
Anti-MKRN1 (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB2501704
Anti-MKRN1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2501717
Anti-MKRN1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2102989
Anti-TRIM27 antibody produced in rabbit, affinity isolated antibody
SAB2700796
Anti-TRIM27 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2700798
Anti-TRIM27 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2700971
Anti-TRIM27 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA048684
Anti-TRIM27 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1403837
Monoclonal Anti-FPRL1 antibody produced in mouse, clone 2G8, purified immunoglobulin, buffered aqueous solution
SAB1402202
Monoclonal Anti-FPRL1 antibody produced in mouse, clone 2D8, purified immunoglobulin, buffered aqueous solution
WH0023608M1
Monoclonal Anti-MKRN1 antibody produced in mouse, clone 4E9, purified immunoglobulin, buffered aqueous solution