EMAIL THIS PAGE TO A FRIEND
Infection and immunity
Characterization of immunological cross-reactivity between enterotoxigenic Escherichia coli heat-stable toxin and human guanylin and uroguanylin.
PMID 24778111
Abstract
Enterotoxigenic Escherichia coli (ETEC) expressing the heat-stable toxin (ST) (human-type [STh] and porcine-type [STp] variants) is among the five most important enteric pathogens in young children living in low- and middle-income countries. ST mediates diarrheal disease through activation of the guanylate cyclase C (GC-C) receptor and is an attractive vaccine target with the potential to confer protection against a wide range of ETEC strains. However, immunological cross-reactivity to the endogenous GC-C ligands guanylin and uroguanylin is a major concern because of the similarities to ST in amino acid sequence, structure, and function. We have investigated the presence of similar epitopes on STh, STp, guanylin, and uroguanylin by analyzing these peptides in eight distinct competitive enzyme-linked immunosorbent assays (ELISAs). A fraction (27%) of a polyclonal anti-STh antibody and an anti-STh monoclonal antibody (MAb) cross-reacted with uroguanylin, the latter with a 73-fold-lower affinity. In contrast, none of the antibodies raised against STp, one polyclonal antibody and three MAbs, cross-reacted with the endogenous peptides. Antibodies raised against guanylin and uroguanylin showed partial cross-reactivity with the ST peptides. Our results demonstrate, for the first time, that immunological cross-reactions between ST and the endogenous peptides can occur. However, the partial nature and low affinity of the observed cross-reactions suggest that the risk of adverse effects from a future ST vaccine may be low. Furthermore, our results suggest that this risk may be reduced or eliminated by basing an ST immunogen on STp or a selectively mutated variant of STh.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A4187 Anti-Goat IgG (whole molecule)–Alkaline Phosphatase antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
A4312 Anti-Mouse IgG (whole molecule)−Alkaline Phosphatase antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
A8025 Anti-Rabbit IgG (whole molecule)–Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution
85191
Glutaraldehyde solution, 1.2 % (w/v) glutaraldehyde in H2O
C5H8O2
49630
Glutaraldehyde solution, technical, ~25% in H2O (2.6 M)
C5H8O2
PHR1359
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
C10H17N3O6S
Y0000517
Glutathione, European Pharmacopoeia (EP) Reference Standard
C10H17N3O6S
G7504
Guanosine 3′,5′-cyclic monophosphate, ≥98% (HPLC), powder
C10H12N5O7P
G6129
Guanosine 3′,5′-cyclic monophosphate sodium salt, ≥99% (HPLC), powder
C10H11N5NaO7P
L5501
L-Lysine, ≥98% (TLC)
C6H14N2O2
62840
L-Lysine, crystallized, ≥98.0% (NT)
C6H14N2O2
23128
L-Lysine, analytical standard
C6H14N2O2
W384704
L-Lysine, ≥97%
C6H14N2O2
62850
L-Lysine solution, purum, 50% in H2O
C6H14N2O2
1551139
Monobasic potassium phosphate, United States Pharmacopeia (USP) Reference Standard
H2KO4P
PHR1330
Monobasic Potassium Phosphate, Pharmaceutical Secondary Standard; Certified Reference Material
H2KO4P
60229
Potassium phosphate monobasic, tested according to Ph.Eur., anhydrous
H2KO4P
U5131
Uroguanylin, ≥90% (HPLC)
C64H102N18O26S4