EMAIL THIS PAGE TO A FRIEND
Acta poloniae pharmaceutica
Material and tableting properties of Azadirachta indica gum with reference to official acacia gum.
PMID 24779199
Abstract
This study determined the material and tableting properties of Azadirachta indica gum (NMG) relative to acacia gum (ACA). The morphological properties were assessed with size and shape factors of aspect ratio, roundness, irregularity and equivalent-circle-diameter. The tableting properties of the gums were determined using compressional characteristics, tensile strength (TS), brittle fracture index (BFI) and crushing-strength-friability/disintegration-time ratio (CSFR/DT). The results suggest that NMG possesses larger, irregular and more elongated particles than ACA. The onset and amount of plastic deformation occurring in NMG was faster and higher, respectively, than in ACA. The result shows that, although ACA tablets were stronger, their tendency to cap/laminate was higher than in NMG tablets. The NMG tablets possess lower DT than those of ACA, while the CSFR/DT result suggests that a better balance exists between the strength and weakness of NMG tablets. The study concluded that NMG can be a useful excipient in tablet formulation.