EMAIL THIS PAGE TO A FRIEND
Molecular pharmaceutics
Hydrogel-based 3D model of patient-derived prostate xenograft tumors suitable for drug screening.
PMID 24779589
Abstract
The lack of effective therapies for bone metastatic prostate cancer (PCa) underscores the need for accurate models of the disease to enable the discovery of new therapeutic targets and to test drug sensitivities of individual tumors. To this end, the patient-derived xenograft (PDX) PCa model using immunocompromised mice was established to model the disease with greater fidelity than is possible with currently employed cell lines grown on tissue culture plastic. However, poorly adherent PDX tumor cells exhibit low viability in standard culture, making it difficult to manipulate these cells for subsequent controlled mechanistic studies. To overcome this challenge, we encapsulated PDX tumor cells within a three-dimensional hyaluronan-based hydrogel and demonstrated that the hydrogel maintains PDX cell viability with continued native androgen receptor expression. Furthermore, a differential sensitivity to docetaxel, a chemotherapeutic drug, was observed as compared to a traditional PCa cell line. These findings underscore the potential impact of this novel 3D PDX PCa model as a diagnostic platform for rapid drug evaluation and ultimately push personalized medicine toward clinical reality.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0001466
Anhydrous Docetaxel, European Pharmacopoeia (EP) Reference Standard
C43H53NO14
MAB1281
Anti-Nuclei Antibody, clone 235-1, 100 µL, Detect Nuclei using this Anti-Nuclei Antibody, clone 235-1 validated for use in IH(P) & IP.
SAB2501669
Anti-CASP3 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA002643
Anti-CASP3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV00021
Anti-CASP3 antibody produced in rabbit, IgG fraction of antiserum
SAB4503291
Anti-CASP3 antibody produced in rabbit, affinity isolated antibody
C9598 Anti-Caspase 3 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
HPA000451
Anti-MKI67 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA001164
Anti-MKI67 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
01885
Docetaxel, purum, ≥97.0% (HPLC)
C43H53NO14
1224551 Docetaxel, United States Pharmacopeia (USP) Reference Standard
C43H53NO14 · 3H2O
469300
Hexamethyldisiloxane, viscosity 0.65 cSt (25 °C)
C6H18OSi2
W326305
L-Cysteine, ≥97%, FG
C3H7NO2S
168149
L-Cysteine, 97%
C3H7NO2S
C7352
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
C3H7NO2S
C5360
L-Cysteine, PharmaGrade, Ajinomoto, from non-animal source, manufactured under appropriate GMP controls for Pharma or Biopharmaceutical production, suitable for cell culture
C3H7NO2S
30089
L-Cysteine, BioUltra, ≥98.5% (RT)
C3H7NO2S
778672
L-Cysteine, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.0%
C3H7NO2S
95437
L-Cysteine, certified reference material, TraceCERT®
C3H7NO2S
WH0000836M2 Monoclonal Anti-CASP3 antibody produced in mouse, clone 4D3, purified immunoglobulin, buffered aqueous solution
C5737 Monoclonal Anti-Caspase 3 antibody produced in mouse, clone 84803.111, purified immunoglobulin, lyophilized powder
WH0004288M1
Monoclonal Anti-MKI67 antibody produced in mouse, clone 7B8, purified immunoglobulin, buffered aqueous solution
AMAB90870
Monoclonal Anti-MKI67 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1234, purified immunoglobulin, buffered aqueous glycerol solution
SAB5300423 Monoclonal Anti-MKI67 antibody produced in mouse, clone 4A1, ascites fluid
SAB5300424 Monoclonal Anti-MKI67 antibody produced in mouse, clone 9C12B2, ascites fluid
SAB5300425 Monoclonal Anti-MKI67 antibody produced in mouse, clone 8D5, ascites fluid
235709
Octamethyltrisiloxane, 98%
C8H24O2Si3