EMAIL THIS PAGE TO A FRIEND
Lung cancer (Amsterdam, Netherlands)
Neoadjuvant chemo-immunotherapy modifies CD4(+)CD25(+) regulatory T cells (Treg) in non-small cell lung cancer (NSCLC) patients.
PMID 24780112
Abstract
Regulatory T cells (Treg) are critical for cancer immune evasion; whereas natural killer (NK) cells are central for effective anti-tumor immunity including antibody-induced cellular cytotoxicity (ADCC). The predictive role of Treg levels for clinical response to chemo-immunotherapy in non-small cell lung cancer (NSCLC) as well as therapy-induced Treg changes remain to be defined. The impact of Treg on NK-mediated cetuximab-dependent cellular cytoxicity was tested in vitro. Frequency and functional activity of Treg was analyzed in 31 NSCLC stage IB-IIIA patients treated by neoadjuvant Cetuximab/Docetaxel/Cisplatin prior to surgery. Data were correlated with clinical outcome variables and Treg tumor infiltration. Treg potently inhibit NK-mediated and cetuximab-induced ADCC in vitro. In addition, a significant correlation between Treg reduction and clinical response was seen. However, the grade of tumor infiltrating Treg in resected tumors did not correlate with peripheral Treg levels. Moreover, Treg levels at diagnosis did not predict clinical response to chemo-immunotherapy. The drop of Treg levels during neoadjuvant chemo-immunotherapy in NSCLC patients significantly correlates with clinical response. However, Treg at diagnosis are not linked to inferior clinical response to chemo-immunotherapy in NSCLC in vivo even though Treg efficiently inhibit ADCC in vitro.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV41764
Anti-GNAS (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV41765
Anti-GNAS (AB2) antibody produced in rabbit, IgG fraction of antiserum
AV42694
Anti-GNAS (AB3) antibody produced in rabbit, IgG fraction of antiserum
SAB2501411
Anti-GNAS antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA018122
Anti-GNAS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2100941
Anti-GNAS antibody produced in rabbit, affinity isolated antibody
HPA027478
Anti-GNAS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1
HPA028386
Anti-GNAS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab2
SAB1400841
Anti-LIPH antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
HPA049079
Anti-LIPH antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2107647
Anti-LIPH antibody produced in rabbit, affinity isolated antibody
HPA021656
Anti-SLC10A3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2105728
Anti-SLC10A3 antibody produced in rabbit, affinity isolated antibody